Skip to Content
Merck
All Photos(5)

Documents

HPA001895

Sigma-Aldrich

Anti-LAMB2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Laminin B1s chain antibody produced in rabbit, Anti-Laminin subunit β-2 precursor antibody produced in rabbit, Anti-S-laminin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DLTDVQDENFNANHALSGLERDRLALNLTLRQLDQHLDLLKHSNFLGAYDSIRHAHSQSAEAERRANTSALAVPSPVSNSASARHRTEALMDAQKEDFNSKHMANQRALGKLSAHTHTLSLTDINELVCGAPG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMB2(3913)

General description

LAMB2 (laminin subunit β 2) is a glycoprotein associated with synaptic basal lamina. It acts as a potent promoter of neurite outgrowth, and essential component of certain kidney membranes and basal laminae in the neuromuscular system. In mamalian neuromuscular system, seven laminin genes have been identified in seven distinct basal laminae (perineurial, endoneurial, terminal Schwann cell, myotendinous junction, synaptic cleft, synaptic fold, and extrajunctional muscle).

Immunogen

Laminin subunit β-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LAMB2 (laminin subunit β 2) is involved with different structural and signaling functions, such as behavioral regulation of nerve, muscle, and Schwann cell. It is expressed in the neural retina for the purpose of the retinal differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85192

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D D Hunter et al.
Neuron, 8(3), 399-413 (1992-03-01)
The development of the neural retina follows a stereotyped time course that begins with an undifferentiated neuroepithelium populated by multipotential progenitor cells and ends with a highly differentiated tissue containing diverse cell types. The identities of the factors that guide
B L Patton
Microscopy research and technique, 51(3), 247-261 (2000-10-31)
The mammalian neuromuscular system expresses seven laminin genes (alpha 1, alpha 2, alpha 4, alpha 5, beta 1, beta 2, and gamma 1), produces seven isoforms of the laminin trimer (laminins 1, 2, 4, 8, 9, 10, and 11), and
D D Hunter et al.
Nature, 338(6212), 229-234 (1989-03-16)
A striking example of topographic specificity in synapse formation is the preferential reinnervation of original synaptic sites on denervated muscle fibres by regenerating motor axons. This specificity is mediated by the basal lamina of the synaptic cleft. A glycoprotein, s-laminin
S E Pors et al.
Human reproduction (Oxford, England), 34(8), 1523-1535 (2019-07-10)
Can a reconstructed ovary using decellularized human ovarian tissue (DCT) support survival of pre-antral stage follicles? We have demonstrated an effective protocol for decellularization of human ovarian tissues and successful recellularization with isolated human ovarian cells and pre-antral follicles. Survivors
M E Durkin et al.
Cytogenetics and cell genetics, 84(3-4), 173-178 (1999-07-07)
The laminin beta2 chain is an important constituent of certain kidney and muscle basement membranes. We have generated a detailed physical map of a 110-kb genomic DNA segment surrounding the human laminin beta2 chain gene (LAMB2) on chromosome 3p21.3-->p21.2, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service