Skip to Content
Merck
All Photos(3)

Key Documents

HPA001600

Sigma-Aldrich

Anti-ELK3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ETS domain-containing protein Elk-3 antibody produced in rabbit, Anti-ETS-related protein ERP antibody produced in rabbit, Anti-ETS-related protein NET antibody produced in rabbit, Anti-SAP-2 antibody produced in rabbit, Anti-SRF accessory protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KNIIKKVIGQKFVYKFVSFPEILKMDPHAVEISRESLLLQDSDCKASPEGREAHKHGLAALRSTSRNEYIHSGLYSSFTINSLQNPPDAFKAIKTEKLEEPPEDSPPVEEVRTVIRFVTNKTDKHVTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELK3(2004)

Immunogen

ETS domain-containing protein Elk-3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ELK3 antibody produced in rabbit is suitable for use to locate ELK3 DNA binding sites on a genome wide scale by a chromatin immunoprecipitation followed by deep sequencing (ChIP-seq) experiment.
Anti-ELK3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

ELK3 (ETS-domain protein) gene encodes a protein belonging to the Ets oncoprotein family. It is localized to human chromosome 12q22–q23. It functions as a transcriptional repressor under basal conditions. It is downregulated in hypoxia and is involved in the regulation of several genes, including c-fos, vascular endothelial growth factor, egr-1 (early growth response protein 1) and HIF-1α (hypoxia-inducible factor 1-α). It forms a ternary complex with Elk-1 and Sap-1 transcription factors and plays a role in early response of quiescent cells to growth factor stimulation. It also functions in angiogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84746

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gross et al.
Oncogene, 27(9), 1333-1341 (2007-08-21)
The ternary complex factor Net/Elk3 is downregulated in hypoxia and participates in the induction by hypoxia of several genes, including c-fos, vascular endothelial growth factor and egr-1. However, the global role of Net in hypoxia remains to be elucidated. We
Qiuyan Zhao et al.
Frontiers in cell and developmental biology, 9, 700192-700192 (2021-08-20)
Rapid progression and metastasis are the major causes of death in patients with pancreatic ductal adenocarcinoma (PDAC). ELK3, a member of the ternary complex factor (TCF), has been associated with the initiation and progression of various cancers. However, the role
Kostyantyn Semenchenko et al.
PloS one, 11(7), e0159531-e0159531 (2016-07-20)
Transcription factors have an important role in cancer but are difficult targets for the development of tumour therapies. These factors include the Ets family, and in this study Elk3 that is activated by Ras oncogene /Erk signalling, and is involved
A Giovane et al.
Genomics, 29(3), 769-772 (1995-10-10)
Net, Elk1, and Sap1 are related members of the Ets oncoprotein family. We show by in situ hybridization on banded chromosomes with specific cDNA probes that their map positions on mouse and human chromosomes (respectively) are net, 10C-D1 and 12q22-q23
E Douglas Robertson et al.
PloS one, 9(11), e113050-e113050 (2014-11-18)
The molecular response to hypoxia is a critical cellular process implicated in cancer, and a target for drug development. The activity of the major player, HIF1α, is regulated at different levels by various factors, including the transcription factor ELK3. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service