Skip to Content
Merck
All Photos(2)

Key Documents

HPA000904

Sigma-Aldrich

Anti-GMPR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GMPR2), transcript variant 2, mRNA, Anti-NM_001002000.1, Anti-guanosine monophosphate reductase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

YPQLSAVMECADAAHGLKGHIISDGGCSCPGDVAKAFGAGADFVMLGGMLAGHSESGGELIERDGKKYKLFYGMSSEMAMKKYAGGVAEYRASEGKTVEVPFKGDVEHTIRDILGGIRSTCTYVGAAKLKELSRRTTFIRVTQQV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GMPR2(51292)

General description

The GMPR2 (guanosine monophosphate reductase 2) gene is mapped to human chromosome 14. The encoded protein is a homotetramer spanning a length of 348 amino acid residues. Each monomer has 11 α-helices and 14 β-sheets. It forms an eight-stranded α/β barrel core. The N and C-terminal sections are attched to this core. Some highly conserved hydrophilic residues of GMPR family are present in this core, making it highly stable.

Immunogen

GMP reductase 2 (Guanosine 5′-monophosphate oxidoreductase 2) (Guanosine monophosphate reductase 2) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene GMPR2 encodes an isozyme of the enzyme guanosine monophosphate reductase (GMPR). It requires NADPH as the coenzyme for its activity. GMPR catalyzes the conversion of nucleoside and nucleotide derivatives of guanine to adenine nucleotides. It maintains the intercellular homeostasis between adenine and guanine. GMPR2 is a novel human GMP reductase and its overexpression can promote the monocytic differentiation of HL-60 leukemia cells. The protein expression is significantly associated with basal-like tumours (BP) at the transcriptome level. Loss of expression of GMPR2 is associated with BP at the protein level and it may have a role in carcinogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70437

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B G Baker et al.
Breast cancer research and treatment, 137(1), 127-137 (2012-12-05)
Basal-like tumours (BP) are a poor prognostic class of breast cancer but remain a biologically and clinically heterogeneous group. We have previously identified two novel genes PPARα (positive) and GMPR2 (negative) whose expression was significantly associated with BP at the
Jixi Li et al.
Journal of molecular biology, 355(5), 980-988 (2005-12-20)
Guanosine monophosphate reductase (GMPR) catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP, and plays a critical role in re-utilization of free intracellular bases and purine nucleosides. Here, we report the first crystal structure of human GMP reductase
Yingfeng Deng et al.
The international journal of biochemistry & cell biology, 34(9), 1035-1050 (2002-05-16)
GMP reductase (EC 1.6.6.8) is the only known metabolic step by which guanine nucleotides can be converted to the pivotal precursor of both adenine and guanine nucleotides. Human GMP reductase has been previously partially purified from erythrocytes and a chromosome
Jia Zhang et al.
Journal of cancer research and clinical oncology, 129(2), 76-83 (2003-04-02)
To identify the biological function of a novel molecule which shows high homology with human guanosine monophosphate reductase (GMPR) and is named GMPR2. GMPR2 cDNA was cloned from the cDNA library of human dendritic cells and was characterized by Bioinformatics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service