Skip to Content
Merck
All Photos(2)

Documents

H9903

Sigma-Aldrich

1-(Heptafluorobutyryl)imidazole

BioReagent, suitable for derivatization

Synonym(s):

1-(Perfluorobutyryl)imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3F7N2O
CAS Number:
Molecular Weight:
264.10
Beilstein:
4488026
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

bp

158-163 °C (lit.)

suitability

suitable for derivatization

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)n1ccnc1

InChI

1S/C7H3F7N2O/c8-5(9,6(10,11)7(12,13)14)4(17)16-2-1-15-3-16/h1-3H

InChI key

MSYHGYDAVLDKCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Heptafluorobutyryl)imidazole, also known as HFBI, is an acylating derivatization reagent. It is a mild amine-group derivatizing agent. HFBI doesn’t produce acidic by-products in reaction, and the unutilized reagent doesn’t damage the chromatographic process. Post derivatization reaction, the HFB (Heptafluorobutyryl) derivatives are analyzed chromatographically.

Application

1-(Heptafluorobutyryl)imidazole is used as a derivatizing agent for analysing Sulphur mustard metabolites. It is suitable to study dispersive derivatization of degradation products.

Biochem/physiol Actions

Mild amine-group derivatizing reagent; non-acidic by-product prevents decomposition and reduces GC column degradation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meehir Palit et al.
Journal of chromatography. A, 1218(32), 5393-5400 (2011-07-09)
A new derivatization and extraction technique termed as dispersive derivatization liquid-liquid extraction (DDLLE) speeds up the analysis process by removing the requirement for drying of the sample. The derivatization process takes place at the interface between the analyte containing aqueous
A Fidder et al.
Archives of toxicology, 74(4-5), 207-214 (2000-08-26)
The development of a procedure for retrospective detection and quantitation of exposure to the arsenical dichloro(2-chlorovinyl)arsine (lewisite; L1) has been initiated. Upon incubation of human blood with [14C]L1 (20 nM-0.2 mM) in vitro, more than 90% of the total radioactivity
Y Luo et al.
Journal of chromatography. B, Biomedical applications, 667(2), 344-348 (1995-05-19)
A method for the monitoring of dihydroetorphine hydrochloride, a powerful anaesthetic and analgesic drug, in biological fluids was developed, involving GC-MS with multiple selected-ion monitoring. Dihydroetorphine was extracted from human blood and urine with dichloromethane and then derivatized with N-heptafluorobutyrylimidazole
Xinxin Li et al.
Colloids and surfaces. B, Biointerfaces, 74(1), 370-374 (2009-09-01)
Patterning of neural stem cells (NSCs) is of great importance for its potential applications in the therapy of nerve injuries. Due to the critical requirements and the great difficulty in NSCs cultivation, developing new methods for NSCs patterning is very
Gas chromatographic determination of 2-ethylhexanol and 2-ethylhexanoic acid as derivatives suitable for electron-capture and nitrogen-phosphorus detection after single reaction with heptafluorobutyrylimidazole.
T Górski et al.
Journal of chromatography, 509(2), 383-389 (1990-06-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service