Skip to Content
Merck
All Photos(1)

Key Documents

H7654

Sigma-Aldrich

Hydroxyguanidine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH5N3O · 1/2H2SO4
CAS Number:
Molecular Weight:
124.11
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (TLC)

form

powder

solubility

water: 25 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

NC(=N)NO.NC(=N)NO.OS(O)(=O)=O

InChI

1S/2CH5N3O.H2O4S/c2*2-1(3)4-5;1-5(2,3)4/h2*5H,(H4,2,3,4);(H2,1,2,3,4)

InChI key

MTGDDPZRXSDPFH-UHFFFAOYSA-N

Biochem/physiol Actions

An early antitumor agent. Oxidation results in release of NO, and formation of other reactive oxygen species, including peroxynitrite and peroxyl radicals. Reacts with NO to form an adduct which is a potent and stable vasodilator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zembowicz, A., et al.
British Journal of Pharmacology, 107, 1007-1007 (1992)
Kamil Brożewicz et al.
European journal of medicinal chemistry, 55, 384-394 (2012-08-16)
Twenty four 1-[2-alkylthio-5-(azol-2 or 5-yl)-4-chlorobenzenesulfonyl]-3-hydroxyguanidines 6a-x have been synthesized in order to evaluate their biological activity. Compounds 6a, 6c, 6d, 6f, 6g, 6i-p, 6r-t, and 6v-x were tested for their in vitro anticancer activity at the US National Cancer Institute. The
Amila Kahvedžić et al.
Journal of medicinal chemistry, 56(2), 451-459 (2012-12-21)
In this paper we report the synthesis of a new family of hydroxyguanidinium aromatic derivatives (4a-g) as potential minor groove binders and cytotoxic agents. Their DNA affinity was evaluated by thermal denaturation experiments using salmon sperm DNA. The antiproliferative effects
T Bill Cai et al.
Current topics in medicinal chemistry, 5(7), 721-736 (2005-08-17)
Nitric oxide (NO) has been implicated in a wide variety of disease states. Both inhibitors and substrates of nitric oxide synthase (NOS) could have great therapeutic potential in the treatment of these diseases. There is considerable pharmacological interest in developing
Alycen Pond Nigro et al.
Archives of biochemistry and biophysics, 500(1), 66-73 (2010-03-30)
Yeast cytochrome c peroxidase was used to construct a model for the reactions catalyzed by the second cycle of nitric oxide synthase. The R48A/W191F mutant introduced a binding site for N-hydroxyguanidine near the distal heme face and removed the redox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service