Skip to Content
Merck
All Photos(1)

Documents

H1917

Sigma-Aldrich

Histone H1 human

Synonym(s):

Histone, Human

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... H1.0(3005)

Looking for similar products? Visit Product Comparison Guide

General description

Histone H1 is part of high-order chromatin structure and is a linker protein associated with nucleosome core particle. Histone H1 is mapped to human chromosome 6. Histone H1 exists in different isoforms. Histone H1 comprises of N-terminal hydrophobic rich region, central globular domain and a C-terminal with intrinsically disordered regions.

Biochem/physiol Actions

The C-terminal regions of histone H1 is essential for binding to chromatin. The placement of H1 is necessary for nucleosome positioning. Histone H1 maintains chromosome architecture and stabilises compaction of nucleosomes during interphase. H1-null mutants display increased rate of chromatin aberration and growth defects. Histone H1 modulates embryogenesis and DNA repair. The H1 histone undergoes posttranslational modification including phosphorylation, methylation and acetylation, each of which carry out different physiological functions. Histone H1 and its variants contribute to the pathogenesis in cancer, Alzheimer′s disease, male infertility and immunity related disorders.

Physical form

Supplied as 1 mg/mL in 20 mM NaPO4, pH 7.0, 0.3 M NaCl, 1 mM EDTA, and 1 mM DTT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H1 histones: current perspectives and challenges
Harshman SW, et al.
Nucleic Acids Research, 41(21), 9593-9609 (2013)
Linker Histone in Diseases
Ye X, et al.
International Journal of Biological Sciences, 13(8), 1008-1018 (2017)
Dynamic placement of the linker histone H1 associated with nucleosome arrangement and gene transcription in early Drosophila embryonic development
Hu J, et al.
Cell Death & Disease, 9(7), 765-765 (2018)
The H1 linker histones: multifunctional proteins beyond the nucleosomal core particle
Hergeth SP and Schneider R
EMBO Reports, 16(11), 1439-1453 (2015)
Histone H1 null vertebrate cells exhibit altered nucleosome architecture
Hashimoto H, et al.
Nucleic Acids Research, 38(11), 3533-3545 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service