Skip to Content
Merck
All Photos(1)

Key Documents

H108

Sigma-Aldrich

Hemicholinium-3

solid, ≥95% (HPLC)

Synonym(s):

2,2′-(4,4′-Biphenylene)bis(2-hydroxy-4,4-dimethylmorpholinium bromide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34Br2N2O4
CAS Number:
Molecular Weight:
574.35
Beilstein:
5704458
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white

solubility

ethanol: 1.3 mg/mL
H2O: 10 mg/mL

SMILES string

[Br-].[Br-].C[N+]1(C)CCOC(O)(C1)c2ccc(cc2)-c3ccc(cc3)C4(O)C[N+](C)(C)CCO4

InChI

1S/C24H34N2O4.2BrH/c1-25(2)13-15-29-23(27,17-25)21-9-5-19(6-10-21)20-7-11-22(12-8-20)24(28)18-26(3,4)14-16-30-24;;/h5-12,27-28H,13-18H2,1-4H3;2*1H/q+2;;/p-2

InChI key

OPYKHUMNFAMIBL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hemicholinium-3 has been used in the preparation of Krebs-HC-3 buffer to study its effects on choline uptake and choline acetyltransferase activity in differentiated human neuroblastoma (SK-N-SH) cells.

Biochem/physiol Actions

Hemicholinium-3 plays a role in blocking the neuronal choline uptake thereby inhibiting acetylcholine synthesis in the brain.

Features and Benefits

This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Yu Kuang et al.
Molecular pharmaceutics, 7(6), 2077-2092 (2010-08-12)
Altered choline (Cho) metabolism in cancerous cells can be used as a basis for molecular imaging with PET using radiolabeled Cho. In this study, the metabolism of tracer Cho was investigated in a woodchuck hepatocellular carcinoma (HCC) cell line (WCH17)
Sebastian A Müller et al.
European journal of nuclear medicine and molecular imaging, 36(9), 1434-1442 (2009-04-09)
Choline derivatives labelled with positron emitters are successfully used for PET imaging of prostate cancer patients. Since little is known about uptake mechanisms, the aim of this study was to characterize choline uptake in prostate cancer cells, also following anti-androgen
Structure-activity aspects of hemicholinium-3 (HC-3) and its analogs and congeners.
J G Cannon
Medicinal research reviews, 14(5), 505-531 (1994-09-01)
Zhuo Li et al.
Biochimica et biophysica acta, 1841(6), 859-867 (2014-03-04)
There is a paucity of information about phosphatidylcholine (PC) biosynthesis in bone formation. Thus, we characterized PC metabolism in both primary human osteoblasts (HOB) and human osteosarcoma MG-63 cells. Our results show that the CDP-choline pathway is the only de
Determination of high-affinity choline uptake (HACU) and choline acetyltransferase (ChAT) activity in the same population of cultured cells
Ray B, et al.
Brain Research, 1297(5), 160-168 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service