Skip to Content
Merck

H0786

Heregulin-β, EGF Domain human

≥80% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Synonym(s):

HRG-β

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

185 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NRG1(3084)

General description

Truncated human sequence (amino acids 178-241) corresponding to the EGF domain and purified as a GST-fusion protein, cleaved with thrombin.

Biochem/physiol Actions

Growth factor ligand for ErbB3 and ErbB4 receptor tyrosine kinases.
Growth factor ligand for ErbB3 and ErbB4 receptor tyrosine kinases. The binding of HRG results in receptor dimerization and receptor trans-autophosphorylation. The phosphorylated receptors recruit cellular signaling proteins, initiating signaling pathways.

Physical form

Solution in phosphate buffered saline containing 30% glycerol.

Analysis Note

Stimulates tyrosine phosphorylation of the ErbB-3/HER-3 receptor.
The biological activity is measured by its ability to activate the c-ErbB3/HER-3 receptor in treated MCF-7 cells.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library