Skip to Content
Merck
All Photos(1)

Key Documents

GW20073F

Sigma-Aldrich

Anti-Complement C3 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AHUS5, Anti-ARMD9, Anti-ASP, Anti-C3a, Anti-C3b, Anti-CPAMD1, Anti-HEL-S-62p

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-20073F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C3(718)

General description

Chicken polyclonal anti-Complement C3 antibody reacts with human complement C3.
Complement C3 plays a central role in the activation of the complement system. It′s processing by C3 convertase is the central reaction in both classical and alternative complement pathways. After activation C3b can bind covalently, via its reactive thioester, to cell surface carbohydrates or immune aggregates. Derived from proteolytic degradation of complement C3, C3a anaphylatoxin is a mediator of local inflammatory process. It induces the contraction of smooth muscle, increases vascular permeability, and causes histamine release from mast cells and basophilic leukocytes.
The complement factor 3 (C3) gene, with 41 exons spanning 41kb on genomic DNA, is mapped to human chromosome 19p13.3. It is the third component of complement and is characterized with α and β chains. It is one of the most abundant complement protein in the serum.

Immunogen

Complement C3 native protein.
Immunogen Sequence: GI # 4557385, sequence 1-1663

Application

Anti-complement C3 antibody produced in chicken has been used in immunoblot analysis.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Chicken polyclonal anti-Complement C3 antibody is used to tag complement C3 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as ELISA and western blotting. It is used as a probe to determine the presence and roles of complement C3 as a component of the complement system.

Biochem/physiol Actions

Complement factor 3 (C3) plays an essential role in the activation of the complement system. C3-convertase (C4b2a) cleaves C3 complement into C3a and C3b. C3b can bind covalently, through its reactive thioester, to cell surface carbohydrates or immune complexes and involve in the self-activation loop of complement activation via the alternate pathway. In addition, C3b also interacts with C3 convertases (C4b2a or C3bBb) to form the C5 convertase, which cleaves complement factor 5 into C5a and C5b. Derived from proteolytic degradation of complement C3, C3a (anaphylatoxin) facilitates the recruitment of inflammatory cells. C3a stimulates smooth muscle contraction, enhances vascular permeability and causes histamine release from mast cells and basophilic leukocytes. Mutation in C3 gene leads to the development of age-related macular degeneration (AMD).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew T Sorbara et al.
Cell host & microbe, 23(5), 644-652 (2018-05-11)
In physiological settings, the complement protein C3 is deposited on all bacteria, including invasive pathogens. However, because experimental host-bacteria systems typically use decomplemented serum to avoid the lytic action of complement, the impact of C3 coating on epithelial cell responses
Rajia Bahri et al.
Frontiers in immunology, 13, 861545-861545 (2022-06-08)
Cutaneous melanoma is one of the most aggressive human malignancies and shows increasing incidence. Mast cells (MCs), long-lived tissue-resident cells that are particularly abundant in human skin where they regulate both innate and adaptive immunity, are associated with melanoma stroma
Michelle Elvington et al.
The Journal of clinical investigation, 127(3), 970-981 (2017-02-14)
An intracellular complement system (ICS) has recently been described in immune and nonimmune human cells. This system can be activated in a convertase-independent manner from intracellular stores of the complement component C3. The source of these stores has not been
Giuseppina Arbore et al.
Nature communications, 9(1), 4186-4186 (2018-10-12)
The induction of human CD4+ Th1 cells requires autocrine stimulation of the complement receptor CD46 in direct crosstalk with a CD4+ T cell-intrinsic NLRP3 inflammasome. However, it is unclear whether human cytotoxic CD8+ T cell (CTL) responses also rely on
Jeffrey R Liddell et al.
Molecular neurodegeneration, 19(1), 14-14 (2024-02-06)
Ferroptosis is a form of regulated cell death characterised by lipid peroxidation as the terminal endpoint and a requirement for iron. Although it protects against cancer and infection, ferroptosis is also implicated in causing neuronal death in degenerative diseases of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service