Skip to Content
Merck

Skip To

GS128

Giemsa Stain, Modified

Synonym(s):

Giemsa Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
41116121
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

pH

6.75-6.95

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Application

When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GS1LGS80GS500
concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

form

solution

form

solution

form

solution

form

solution

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use


Still not finding the right product?

Explore all of our products under Giemsa Stain, Modified


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters