Skip to Content
Merck
All Photos(1)

Key Documents

GERPN999

ECL Plex Western Blotting Combination Pack (Cy®3, Cy5, Hybond-LFP)

Cytiva RPN999, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

description

ECL Plex Goat-anti-mouse IgG-Cy3, 150 ug: Pink ECL Plex Goat-anti-rabbit IgG-Cy5, 150 ug: Blue ECL Plex fluorescent rainbow markers: Brownish-red Hybond-LFP 20 x 20 cm, 3 sheets: White

feature

wetted part: no

packaging

pack of 1 kit

manufacturer/tradename

Cytiva RPN999

storage condition

protect from light

shipped in

dry ice

storage temp.

−20°C

General description

ECL Plex Western Blotting Combination Pack (Cy3, Cy5, Hybond-LFP). Includes the following, sufficient for at least 1000 cm2 of membrane:
ECL Plex Goat-α-mouse IgG-Cy3, 150 μg PA43009
ECL Plex Goat-α-rabbit IgG-Cy5, 150 μg PA45011
ECL Plex Fluorescent Rainbow Markers, 120 μL RPN850E
Hybond® ECL, 10 × 10 cm, 10 sheets or Hybond-LFP, 20 × 20 cm, 3 sheets RPN2020LFP3
Protocol.

Features and Benefits

  • Complete system optimized for fluorescent Western blotting.
  • Improved formulation of Cy3 conjugates for even lower background.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 2 to 8 °C (ECL Plex CyDye conjugated, ECL Plex Fluorescent Rainbow Markers, -15C to -30C, Hybond ECL membrane, dry in sealed bag at room temperature)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact [email protected].

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva
Cy3 is a trademark of Cytiva
ECL Plex is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Hybond-LFP is a trademark of Cytiva
Rainbow is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service