Skip to Content
Merck
All Photos(1)

Documents

GE17-5478-14

HiScreen Capto L

Cytiva 17-5478-14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

recombinant protein L (E. coli), mammalian free

packaging

pkg of 4.7 mL

manufacturer/tradename

Cytiva 17-5478-14

storage condition

(20% Ehtanol)

parameter

44 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

rigid, highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

working range

2-10(pH interval where the medium can be operated without significant change in function.)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Features and Benefits

  • Suitable for the capture of a wide range of antibody fragments such as Fabs, dAbs, and single-chain fragment variable (scFv) .
  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of resin.
  • Enables a platform approach to antibody fragment purification.
  • Excellent productivity, process economy, and process design flexibility due to rigid matrix.
  • Hydrophilic base matrix ensures low levels of non-specific binding and low levels of host cell-derived impurities in the elution pool.
  • Fulfills industrial demands for security of supply, robust performance, and regulatory support.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiScreen is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service