GBL665384
Grace Bio-Labs silicone isolator sheet material
thickness 2.5 mm, (Adhesive on one side)
Synonym(s):
adhesive isolator, silicone blocker
All Photos(2)
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
Recommended Products
material
red silicon
sterility
non-sterile
feature
(Adhesive on one side)
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 665384
L × W
180 mm × 130 mm
thickness
2.5 mm
Related Categories
General description
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available in CultureWell “water-clear” silicone, which is highly recommended for sensitive cell culture applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)