GBL623503
Grace Bio-Labs SecureSeal™ hybridization chambers
wells, 1, well L × W × depth from septum to bottom of cone 22 mm × 22 mm × 0.8 mm, port diam. 1.5 mm
Synonym(s):
microarray chambers for slides, single use silicone-gasketed chamber
About This Item
Recommended Products
material
clear polycarbonate backing (0.25mm thickness)
red silicon chamber
sterility
non-sterile
feature
wells 1
packaging
pack of 50 ea
manufacturer/tradename
Grace Bio-Labs 623503
backing thickness
0.25 mm
external L × W
25 mm × 25 mm
port diam.
1.5 mm
well L × W × depth
22 mm × 22 mm × 0.8 mm
wells
1
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SecureSeal chambers form removable, peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. The chamber design minimizes friction, promotes reagent mixing, and facilitates uniform hybridization. Sealable access ports in the chamber surface allow for the addition and removal of reactants. SecureSeal adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating. Adhesive Seal Tabs create leak-proof chambers that are temperature resistant. Each package includes illustrated instructions for use and 120 adhesive port seals. Additional seal tabs are sold separately GBL629200). Press fit tubing connectors provide affordable and easy means to connect SecureSeal Hybridization Chambers to pumps and other devices for fluidic exchange.
Note: If you are using Cy5 of Alexa Fluor 647 direct-labeled DNA probes, please refer to "fluorescent friendly" chambers
Application
Protein and DNA Microarrays
Immunocytochemistry
DNA Amplification
Neurophysiology
Reaction kinetics
Rapid microfluidic prototyping
Nucleic Acid Programmable Protein Arrays
Fluorescence Resonance Energy Transfer (FRET)
Single molecule fluorescence microscopy
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service