GBL505012
Grace Bio-Labs ONCYTE® NOVA™ nitrocellulose film slides
12- 6 mm x 6 mm NC pad per slide, plain glass
Synonym(s):
nitrocellulose slides, protein microarrays
All Photos(2)
About This Item
UNSPSC Code:
41122600
NACRES:
NB.15
Recommended Products
material
nitrocellulose (on glass)
packaging
pack of 20 ea
manufacturer/tradename
Grace Bio-Labs 505012
external size
25 mm × 75 mm
binding type
medium binding surface
Related Categories
General description
Grace Bio-Labs ONCYTE® NOVA™ nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Legal Information
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)