Skip to Content
Merck
All Photos(1)

Documents

G6918

Supelco

Glucose Standard Solution

1 mg/mL

Synonym(s):

Glucose Reference Standard Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

biological source

maize

Quality Level

description

Contains 0.1% (w/v) benzoic acid

form

liquid

concentration

1 mg/mL

application(s)

clinical testing
detection

format

single component solution

storage temp.

2-8°C

Application

The amount of glycogen in mouse muscle and liver tiβues was indirectly measured by analyzing the level of glycogen-derived glucose by an enzymatic-colorimetric glucose aβay kit using glucose standard solution to determine glycogen levels.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny Fäldt et al.
Endocrinology, 145(6), 2680-2686 (2004-02-28)
IL-6 is produced and released in large amounts from skeletal muscle during prolonged exercise in both mice and humans, but there are few data indicating the biological significance of this. IL-6 exerts metabolic effects such as stimulating energy expenditure and
Jan Hubert et al.
Microbial ecology, 77(4), 1048-1066 (2018-11-23)
Interactions with microorganisms might enable house dust mites (HDMs) to derive nutrients from difficult-to-digest structural proteins and to flourish in human houses. We tested this hypothesis by investigating the effects of changes in the mite culture growth and population of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service