Skip to Content
Merck
All Photos(4)

Key Documents

G5422

Sigma-Aldrich

β-Glycerophosphate disodium salt hydrate

≤1.0 mol % L-α-isomer

Synonym(s):

BGP, Glycerol 2-phosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2CHOP(O)(ONa)2 · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

form

powder

impurities

≤0.1% Inorganic phosphorus
≤1.0 mol % L-α-isomer

mp

102-104 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

O.[Na+].[Na+].OCC(CO)OP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

ROPZSVKNEIIIDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Glycerophosphate is a classical serine-threonine phosphatase inhibitor used in kinase reaction buffers. BGP is often used in combination with other phosphatase/protease inhibitors for broad spectrum inhibition. It functions as an organic phosphate donor and has been used in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. BGP is also used to buffer M17 media for Lactococcus culture in recombinant protein expression.
β-glycerophosphate is used in homegenization solutions for muscles extracts following fixation. [1]
Initiates differentiation of bone-marrow-derived stem cells embedded in a natural collagen/chitosan gel designed for tissue engineering.

Quality

Low inorganic phosphate content. Mixtures of α- and β-glycerophosphate have historically been used in microbiological media but high levels of inorganic (free) phosphate has been associated with the irregular growth of bacteria and unwanted precipitates.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Morten Donsmark et al.
The Journal of physiology, 550(Pt 3), 845-854 (2003-06-10)
Intramuscular triacylglycerol is an important energy store and is also related to insulin resistance. The mobilization of fatty acids from this pool is probably regulated by hormone-sensitive lipase (HSL), which has recently been shown to exist in muscle and to
Yang An et al.
Biochemical and biophysical research communications, 516(2), 551-557 (2019-06-27)
Developing methods for regenerating bones and uncovering the molecular mechanism underlying bone formation have great significance to human health. In the last decade, people have been using adipose-derived stem cells (ADSCs), that are capable of multilineage differentiation, to reconstruct defected
J Douglas et al.
The Journal of applied bacteriology, 56(2), 321-326 (1984-04-01)
An easily prepared medium, originally designed for the cultivation of lactic phages, has been found to have much wider application. Experience in its use over a ten year period with a range of physiologically diverse bacteria, for teaching and research
Gretl Hendrickx et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 36(2), 369-384 (2020-11-13)
The skeleton is a dynamic tissue continuously adapting to mechanical stimuli. Although matrix-embedded osteocytes are considered as the key mechanoresponsive bone cells, all other skeletal cell types are principally exposed to macroenvironmental and microenvironmental mechanical influences that could potentially affect
Lucas R Brun et al.
SpringerPlus, 3, 676-676 (2014-12-19)
The aim of this study was to evaluate the effect of strontium ranelate (SrR) on bone mineral density (BMD) and bone turnover markers after 1 year of treatment. Additionally, the effect of SrR in bisphosphonate-naïve patients (BP-naïve) compared to patients previously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service