Skip to Content
Merck
All Photos(1)

Key Documents

G3882

Sigma-Aldrich

Gly-Gly-Gly-Gly

≥98% (TLC)

Synonym(s):

Glycyl-glycyl-glycyl-glycine, Tetraglycine, Triglycyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CO(NHCH2CO)3OH
CAS Number:
Molecular Weight:
246.22
Beilstein:
1715387
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Gly-Gly-Gly-Gly,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

mp

300 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

NCC(=O)NCC(=O)NCC(=O)NCC(O)=O

InChI

1S/C8H14N4O5/c9-1-5(13)10-2-6(14)11-3-7(15)12-4-8(16)17/h1-4,9H2,(H,10,13)(H,11,14)(H,12,15)(H,16,17)

InChI key

QMOQBVOBWVNSNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Gly-Gly-Gly

Biochem/physiol Actions

Tetraglycine is used with copper (Cu-II) to study mechanisms of hydrogen peroxide/bicarbonate free radical production and effect in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Barthels et al.
ChemMedChem, 15(10), 839-850 (2020-03-03)
Staphylococcus aureus is one of the most frequent causes of nosocomial and community-acquired infections, with drug-resistant strains being responsible for tens of thousands of deaths per year. S. aureus sortase A inhibitors are designed to interfere with virulence determinants. We
Takeshi Nagata et al.
The Journal of chemical physics, 134(3), 034110-034110 (2011-01-26)
The gradient for the fragment molecular orbital (FMO) method interfaced with effective fragment potentials (EFP), denoted by FMO∕EFP, was developed and applied to polypeptides solvated in water. The structures of neutral and zwitterionic tetraglycine immersed in water layers of 2.0
Travis Cooper et al.
Journal of the American Society for Mass Spectrometry, 17(12), 1654-1664 (2006-08-29)
Extensive 13C, 15N, and 2H labeling of tetraglycine was used to investigate the b3+ --> a3* reaction during low-energy collision-induced dissociation (CID) in a quadrupole ion-trap mass spectrometer. The patterns observed with respect to the retention or elimination of the
J A Vazquez et al.
The American journal of physiology, 249(5 Pt 1), G563-G566 (1985-11-01)
Jejunal disappearance rates of glycine (a model for neutral amino acid absorption), triglycine (a model for peptide transport), and tetraglycine (a model for brush-border membrane hydrolysis) were investigated by an in situ perfusion technique before and after 2 wk of
Bojidarka B Ivanova et al.
Biopolymers, 82(6), 587-596 (2006-03-23)
As part of an investigation on the coordination ability of peptides, structural analyses of the solid di-, tri, and tetrapeptides glycyl-glycine (GG), glycyl-glycyl-glycine (GGG), glycyl-glycyl-glycyl-glycine (GGGG), and their protonated hydrochlorides glycyl-glycine.HCl (GGH), glycyl-glycyl-glycine.HCl (GGGH), and glycyl-glycyl-glycyl-glycine.HCl (GGGGH) have been carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service