Skip to Content
Merck
All Photos(1)

Key Documents

G2752

Sigma-Aldrich

Gly-Phe

≥97.0% (TLC)

Synonym(s):

Glycyl-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH(COOH)CH2C6H5
CAS Number:
Molecular Weight:
222.24
Beilstein:
2279318
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Gly-Phe,

Assay

≥97.0% (TLC)

Quality Level

form

crystalline

color

white

mp

264 °C

storage temp.

−20°C

SMILES string

NCC(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C11H14N2O3/c12-7-10(14)13-9(11(15)16)6-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,14)(H,15,16)/t9-/m0/s1

InChI key

JBCLFWXMTIKCCB-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuniyo Inouye et al.
Bioscience, biotechnology, and biochemistry, 71(8), 2083-2086 (2007-08-11)
The aim of this study was to improve the performance of affinity gels containing glycyl-D-phenylalanine (Gly-D-Phe) as a ligand to thermolysin. Gly-D-Phe was immobilized to the resin through spacers of varying chain lengths. The resulting affinity gels had spacer chain
Roderick Y H Lim et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(25), 9512-9517 (2006-06-14)
Natively unfolded phenylalanine-glycine (FG)-repeat domains are alleged to form the physical constituents of the selective barrier-gate in nuclear pore complexes during nucleocytoplasmic transport. Presently, the biophysical mechanism behind the selective gate remains speculative because of a lack of information regarding
Susan Weng Larsen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 22(5), 399-408 (2004-07-22)
Oil-based depot formulations may constitute a future delivery method for small peptides. Thus, a requirement is attainment of sufficient oil solubility for such active compounds. A model dipeptide (Gly-Phe) has been converted into lipophilic prodrugs by esterification at the C-terminal
T Takarada et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(21), 3906-3913 (2000-12-29)
Oligopeptides are efficiently hydrolyzed by Ce(IV) to the corresponding amino acids under mild conditions. The pseudo first-order rate constants for the hydrolysis of H-Gly-Phe-OH and H-Gly-Gly-OH at pH 7.0 and 50 degrees C are 3.5 x 10(-1) and 2.8 x
Lanthanide ion-mediated peptide hydrolysis.
M Komiyama
Metal ions in biological systems, 38, 25-41 (2001-02-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service