Skip to Content
Merck
All Photos(1)

Documents

G2626

Sigma-Aldrich

L-Glutamic Dehydrogenase from bovine liver

Type II, 50% glycerol solution, ≥35 units/mg protein

Synonym(s):

L-GLDH, L-Glutamate:NAD[P]+ Oxidoreductase (deaminating), Glutamate Dehydrogenase from bovine liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine liver

type

Type II

form

glycerol solution (50%)

specific activity

≥35 units/mg protein

mol wt

310-350 kDa

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-glutamic dehydrogenase was used to catalyze the conversion of isocitrate into α-ketoglutarate and carbon dioxide.

Biochem/physiol Actions

Mammalian forms of this enzyme, including this bovine form, can use either NADP(H) or NAD(H) as coenzymes. L-glutamic dehydrogenase plays a unique role in mammalian metabolism. The reverse reaction catalyzed by this enzyme is the only pathway by which ammonia can become bound to the α-carbon atom of an α-carboxylic acid and thus, is the only source of de novo amino acid synthesis in mammalian species.

The bovine enzyme is characterized by three sets of properties:
  • It has a reversible concentration-dependent association, producing higher molecular weight forms.
  • Forms tight enzyme-reduced coenzyme-substrate ternary complexes whose rates of dissociation modulate the steady-state reaction rates.
  • Exhibits a wide variety of effects from the binding of any of a number of nucleotide modifiers.

L-glutamic dehydrogenase catalyzes the conversion of glutamate to α-ketoglutarate.

Unit Definition

One unit will reduce 1.0 μmole of α-ketoglutarate to L-glutamate per min at pH 7.3 at 25 °C, in the presence of ammonium ions.

Physical form

50% glycerol solution

Analysis Note

Protein determined by biuret.

substrate

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing the role of the residues in the active site of the transaminase from Thermobaculum terrenum.
Bezsudnova, et al.
PLoS ONE, 16, e0255098-e0255098 (2021)
Vedrana Montana et al.
Neurochemical research, 46(10), 2551-2579 (2021-06-01)
Astrocytes have a prominent role in metabolic homeostasis of the brain and can signal to adjacent neurons by releasing glutamate via a process of regulated exocytosis. Astrocytes synthesize glutamate de novo owing to the pyruvate entry to the citric/tricarboxylic acid
Mariagrazia Grimaldi et al.
Human molecular genetics, 26(18), 3453-3465 (2017-09-16)
Congenital hyperinsulinism/hyperammonemia (HI/HA) syndrome gives rise to unregulated protein-induced insulin secretion from pancreatic beta-cells, fasting hypoglycemia and elevated plasma ammonia levels. Mutations associated with HI/HA were identified in the Glud1 gene, encoding for glutamate dehydrogenase (GDH). We aimed at identifying
S Vesce et al.
FEBS letters, 411(1), 107-109 (1997-07-07)
The mechanisms of HIV-1 neurotoxicity remain still undefined although the induction of signalling events and a modest inhibition of glutamate uptake induced by the envelope glycoprotein, gp120, have called attention to astrocytes. Here we demonstrate that the levels at which
Anna K Junk et al.
Molecular vision, 16, 1286-1291 (2010-07-29)
To determine whether activity of carbohydrate metabolism enzymes (aldolase, pyruvate kinase, isocitrate dehydrogenase, and malate dehydrogenase) are altered in the glaucomatous trabecular meshwork (TM) compared to controls. Tissue specimens were obtained from trabeculectomy (n=45 open angle glaucoma; Caucasian, average age

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service