Skip to Content

G2018

γ-Globulins from rabbit

≥99% (agarose gel electrophoresis)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
EC Number:
232-706-1
NACRES:
NA.61
UNSPSC Code:
12352202
Form:
powder
Assay:
≥99% (agarose gel electrophoresis)
Biological source:
rabbit
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

assay

≥99% (agarose gel electrophoresis)

form

powder

impurities

≤5% NaCl

solubility

0.9% NaCl: soluble 50 mg/mL

storage temp.

−20°C

General description

γ-Globulin molecules is made up of four polypeptide chains, of which two have molecular weights of about 50,000 and two of about 20,000. These chains are cross-linked by disulfide bonds.

Application

γ-Globulins was used in the isolation and rearrangement of simple isoimides (iminoanhydrides).

Preparation Note

Prepared from plasma


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library