Skip to Content
Merck
All Photos(1)

Key Documents

G1660

Sigma-Aldrich

Galanthamine hydrobromide from Lycoris sp.

≥94% (HPLC)

Synonym(s):

Galantamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3 · HBr
CAS Number:
Molecular Weight:
368.27
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

biological source

plant (Ungeria victoris)

Quality Level

Assay

≥94% (HPLC)

form

powder

mp

270 °C

solubility

DMSO: soluble 10 mg/mL, clear, colorless
water: soluble 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Br[H].COc1ccc2CN(C)CC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24

InChI

1S/C17H21NO3.BrH/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17;/h3-6,12,14,19H,7-10H2,1-2H3;1H/t12-,14-,17-;/m0./s1

InChI key

QORVDGQLPPAFRS-XPSHAMGMSA-N

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

General description

Galanthamine hydrobromide is a derivative of galanthamine, which is an anticholinestrase alkaloid derived from genus Lycoris (Amaryllidaceae) or snowdrop. This compound is used for the treatment of Alzheimer′s disease (AD).

Application

Galanthamine hydrobromide from Lycoris sp. has been used-
  • as an AChE (acetylcholinesterase) inhibitor and positive control in acetylcholinesterase assay performed to determine the activity of AChE
  • as a positive control in the screening of AChE inhibitors
  • as a positive control in AChE inhibitory assay performed to determine the AChE-inhibitory activities of pyrithione and related sulfur-containing pyridine N-oxides

Biochem/physiol Actions

Cholinesterase inhibitor; reverses scopolamine-induced amnesia; antimyasthenic.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Hao et al.
Molecules (Basel, Switzerland), 18(3), 2458-2468 (2013-02-27)
Phytochemical investigation of the 80% ethanol extract of the bulbs of Lycoris radiata resulted in the isolation of five new Amaryllidaceae alkaloids: (+)-5,6-dehydrolycorine (1), (+)-3α,6β-diacetyl-bulbispermine (2), (+)-3α-hydroxy-6β-acetyl- bulbispermine (3), (+)-8,9-methylenedioxylhomolycorine-N-oxide (5), and 5,6-dihydro-5- methyl-2-hydroxyphenanthridine (7), together with two known compounds
Muhammad Ayaz et al.
BMC complementary and alternative medicine, 14, 145-145 (2014-06-03)
We investigated Polygonum hydropiper L. (P. hydropiper) for phenolic contents, antioxidant, anticholinesterase activities, in an attempt to rationalize its use in neurological disorders. Plant crude extract (Ph.Cr), its subsequent fractions: n-hexane (Ph.Hex), chloroform (Ph.Chf), ethyl acetate (Ph.EtAc), n-Butanol (Ph.Bt), aqueous
Petra Krejčová et al.
Journal of ethnopharmacology, 154(1), 176-182 (2014-04-12)
Persian shallot (Allium stipitatum) is a bulbous plant native to Turkey, Iran and Central Asia. It is frequently used in folk medicine for the treatment of a variety of disorders, including inflammation and stress. Antiinflammatory and neurological activities of pyrithione
Lars Sonde et al.
PloS one, 8(2), e57161-e57161 (2013-02-26)
It is largely unknown how the medical treatment of patients diagnosed with dementia is followed up in primary care. Therefore, we studied patient medical records from two dementia clinics and from the referring primary care centres. A retrospective study of
Carina Wattmo et al.
BMC neurology, 12, 134-134 (2012-11-07)
Activities of daily living (ADL) are an essential part of the diagnostic criteria for Alzheimer's disease (AD). A decline in ADL affects independent living and has a strong negative impact on caregiver burden. Functional response to cholinesterase inhibitor (ChEI) treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service