Recommended Products
Quality Level
technique(s)
affinity chromatography: suitable
matrix active group
phase
application(s)
life science and biopharma
compatibility
Cytiva
Looking for similar products? Visit Product Comparison Guide
Application
Sephadex® G-15 has been used in the total antioxidant status (TAS) assay. It has also been used to desalt fractions of sea cucumber Holothuria hilla to isolate polysaccharides.
Sephadex® G-15 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
Fractionation Range (MW)
Globular Proteins: <1500
Dextrans: <1500
Fractionation Range (MW)
Globular Proteins: <1500
Dextrans: <1500
Other Notes
G15120-100G′s updated product number is GE17-0020-01
Legal Information
Sephadex is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Analysis of N(?) -Ethyllysine in Human Plasma Proteins by Gas Chromatography-Negative Ion Chemical Ionization/Mass Spectrometry as a Biomarker for Exposure to Acetaldehyde and Alcohol.
Mabuchi, R., et al.
Alcoholism, Clinical and Experimental Research (2012)
Nadezhda E Ustyuzhanina et al.
Marine drugs, 18(11) (2020-11-01)
Fucosylated chondroitin sulfates (FCSs) PC and HH were isolated from the sea cucumbers Paracaudina chilensis and Holothuria hilla, respectively. The purification of the polysaccharides was carried out by anion-exchange chromatography on a DEAE-Sephacel column. The structural characterization of the polysaccharides
Juan Bruzzo et al.
Journal of cancer research and clinical oncology, 136(10), 1605-1615 (2010-08-12)
The phenomenon of hormesis is characterized by a biphasic dose-response, exhibiting opposite effects in the low- and high-dose zones. In this study, we explored the possibility that the hormesis concept may describe the interactions between two tumors implanted in a
L Ramió-Lluch et al.
Animal reproduction science, 115(1-4), 189-200 (2009-01-23)
This study was performed to test the effects of filtration through several chromatographic resins on the semen quality parameters (percentages of viability, altered acrosomes and morphological abnormalities, motion characteristics and the response to the Osmotic Resistance Test) of boar ejaculates
Qiang Luo et al.
Journal of medicinal food, 14(12), 1609-1616 (2011-09-01)
Crude water-soluble Chinese truffle Tuber indicum polysaccharide (TIP) was extracted from the fruiting bodies with water and then successively purified by DEAE-cellulose 52 and Sephadex G-100 column chromatography, yielding two major polysaccharide fractions: TIP1-1 and TIP2-1. High-performance gel permeation chromatography
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service