Skip to Content
Merck
All Photos(1)

Documents

G1276

Sigma-Aldrich

Gastrin I rat

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H128N22O31S2
CAS Number:
Molecular Weight:
2126.28
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

rat

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)C(Cc3ccc(O)cc3)NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCSC)NC(=O)C4CCCN4C(=O)C5CCCN5C(=O)C(CCCNC(N)=N)NC(=O)C6CCC(=O)N6)C(=O)NC(CC(O)=O)C(=O)NC(Cc7ccccc7)C(N)=O

InChI

1S/C94H128N22O31S2/c1-48(79(133)113-65(43-50-19-21-52(117)22-20-50)80(134)100-47-71(119)103-66(44-51-46-99-54-15-8-7-14-53(51)54)89(143)109-61(35-40-148-2)88(142)114-67(45-77(130)131)90(144)112-64(78(95)132)42-49-12-5-4-6-13-49)101-81(135)56(24-30-72(120)121)104-83(137)57(25-31-73(122)123)105-84(138)58(26-32-74(124)125)106-85(139)59(27-33-75(126)127)107-86(140)60(28-34-76(128)129)108-87(141)62(36-41-149-3)110-91(145)68-17-10-38-115(68)93(147)69-18-11-39-116(69)92(146)63(16-9-37-98-94(96)97)111-82(136)55-23-29-70(118)102-55/h4-8,12-15,19-22,46,48,55-69,99,117H,9-11,16-18,23-45,47H2,1-3H3,(H2,95,132)(H,100,134)(H,101,135)(H,102,118)(H,103,119)(H,104,137)(H,105,138)(H,106,139)(H,107,140)(H,108,141)(H,109,143)(H,110,145)(H,111,136)(H,112,144)(H,113,133)(H,114,142)(H,120,121)(H,122,123)(H,124,125)(H,126,127)(H,128,129)(H,130,131)(H4,96,97,98)

InChI key

FUTWNIPXZKAJHO-UHFFFAOYSA-N

Gene Information

rat ... Gast(25320)

Amino Acid Sequence

Glp-Arg-Pro-Pro-Met-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2

General description

Gastrin is a pyloric antral hormone, which is a small biologically active peptide thatis synthesized as a precursor. It is found to be produced in antral G cells.

Application

Gastrin I has been used in a study for gastrin stimulation in rats to determine the expression of histidine decarboxylase and histamine metabolism in gastric oxyntic mucosa during conditions of hypergastrinemia and carcinoid tumor. It has also been used for preabsorption of anti-gastrin antibody prior to immunohistochemical assay in rats.

Biochem/physiol Actions

Gastrin plays a crucial role in the maintenance of architecture and function of oxyntic mucosa of stomach, which is involved in the secretion of acid. It also triggers the secretion of acid into the gastric lumen. Continuous increased plasma levels of gastrin or hypergastrinemia, may be involved in gastrointestinal carcinogenesis, especially if it is accompanied with inflammation. The release of this peptide is suppressed during distention, which is a result of omatostatin and intrinsic cholinergic pathways. In rats, this peptide maintains gastric mucosal integrity through the up-regulation of cyclooxygenase-2 which is, in part, mediated by heparin-binding epidermal growth factor-like growth factor (HB-EGF).

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinjuan Fu et al.
Molecular medicine (Cambridge, Mass.), 27(1), 90-90 (2021-08-21)
It is known that increased gastrin concentration is negatively correlated with cardiovascular mortality, and plasma gastrin levels are increased in patients after myocardial infarction (MI). However, whether gastrin can play a protective role in MI remains unknown. Adult C57BL/6 mice
L Kölby et al.
Endocrinology, 137(10), 4435-4442 (1996-10-01)
Histamine is an important stimulator of gastric acid secretion. In experimental animals, inhibition of acid secretion by long term histamine2 receptor blockade causes hypergastrinemia, proliferation of enterochromaffin-like (ECL) cells, and formation of histamine-producing gastric carcinoids. The aim of this study
B Hunyady et al.
Endocrinology, 139(10), 4404-4415 (1998-09-29)
Gastrin and histamine both potently stimulate secretion of acid into the gastric lumen. How these agents interact and how their release is controlled is poorly understood. Therefore, we decided to look for histamine in the antral portion of the rat
A Varro et al.
The Journal of biological chemistry, 265(35), 21476-21481 (1990-12-15)
Post-translational processing of the precursor for rat gastrin yields products that include peptides phosphorylated at Ser96, amidated at Phe92, and sulfated at Tyr87 or Tyr103. The phosphorylation site is immediately adjacent to the processing point that gives rise to the
Yong-Yu Li
World journal of gastroenterology, 9(1), 129-133 (2003-01-01)
To investigate the intragastric mechanisms for regulation of gastric neuroendocrine functions during gastric distention in isolated vascularly perfused rat stomach. Isolated vascularly perfused rat stomach was prepared, then the gastric lumen was distended with either 5,10 or 15 ml pH7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service