Skip to Content
Merck
All Photos(3)

Documents

G0500

Sigma-Aldrich

D-(+)-Galactosamine hydrochloride

≥99% (HPLC)

Synonym(s):

2-Amino-2-deoxy-D-galactopyranose hydrochloride, D-Chondrosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
3697825
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

shark (cartilage)

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.5% Glucosamine (HPAE)

color

white to off-white

mp

172-180  °C

solubility

H2O: 50 mg/mL, clear to slightly hazy, colorless to very faintly yellow

storage temp.

room temp

SMILES string

Cl.N[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)CO

InChI

1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4+,5+,6-;/m0./s1

InChI key

CBOJBBMQJBVCMW-NQZVPSPJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-galactosamine (d-GalN) is a specific hepatotoxic agent metabolized particularly in hepatocytes. It is a 6-carbon amino sugar derived from galactose.

Application

D-(+)-Galactosamine (D-chondrosamine) has been used with lipopolysaccharides (LPS) to induce models of acute hepatic failure (LPS/D-GalN-induced liver injury, hepatitis) for therapeutic research to find new drugs.
D-(+)-Galactosamine hydrochloride has been used:
  • for the surface binding of mannan-binding lectin (MBL) to modified mica surfaces
  • in sterile phosphate buffer saline (PBS) before the intraperitoneal injection
  • in mice for the generation of primary bone marrow-derived macrophages (BMDMs)

Biochem/physiol Actions

Galactosamine (Gal) induces hepatocyte death both by necrosis and apoptosis. It prevents the production of liver RNA via the production of uridine diphosphate hexosamines. D-galactosamine lowers the intracellular pool of uracil nucleotides and thereby prevents the production of RNA and proteins.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yating Li et al.
Applied microbiology and biotechnology, 103(1), 375-393 (2018-10-23)
Acute liver failure is a drastic, unpredictable clinical syndrome with high mortality. Various preventive and adjuvant therapies based on modulating the gut flora have been proposed for hepatic injury. We aimed to explore the preventive and therapeutic effects of Bifidobacterium
Kaicen Wang et al.
mSphere, 5(1) (2020-01-31)
Acute liver failure is a severe liver disorder that poses considerable global challenges. Previous studies on Bifidobacterium longum R0175 have mainly focused on its psychotropic functions. The current research focused on the protective efficacy of B. longum R0175 against acute
Aoxiang Zhuge et al.
Applied microbiology and biotechnology, 104(17), 7437-7455 (2020-07-16)
Acute liver failure is a clinical emergency associated with high mortality. Accumulating evidence indicates that gut microbiota participates in the progression of liver injury, and preventive therapies based on altering gut microbiota are of great interest. Previous studies demonstrated that
Xing Lin et al.
Biological & pharmaceutical bulletin, 37(4), 625-632 (2014-05-13)
This study examined the effect of genistein from Hydrocotyle sibthorpioides on lipopolysaccharide (LPS)/D-galactosamine (D-GalN)-induced acute hepatic failure. Compared to the model control, genistein treatment significantly protected against LPS/D-GalN-induced liver injury, as evidenced by the decrease in serum alanine and aspartate
Shan-Shan Li et al.
Molecular immunology, 101, 10-18 (2018-06-01)
Acute liver failure is a devastating clinical syndrome with extremely terrible inflammation reaction, which is still lack of effective treatment in clinic. Suppressor of Cytokine Signaling 1 protein is inducible intracellular negative regulator of Janus kinases (JAK)/signal transducers and activators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service