Skip to Content
Merck
All Photos(2)

Documents

G0282

Sigma-Aldrich

Granulocyte-Macrophage Colony-Stimulating Factor from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

GM-CSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Granulocyte-Macrophage Colony-Stimulating Factor from mouse, GM-CSF, from mouse, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

≤0.2 ng/mL EC50 (corresponds to ≥5 × 106 units/mg)

quality

endotoxin tested

mol wt

~15 kDa (125 amino acids including N-terminal methionine)

packaging

pkg of 5 and 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/mg

color

white

solubility

water: soluble, clear, colorless

UniProt accession no.

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Granulocyte-macrophage colony stimulating factor (GM-CSF) is a growth and differentiation factor for cells in the granulocyte, macrophage and eosinophil lineage. GM-CSF stimulates colony formation from pluripotential progenitor cells at extremely low concentrations and is an essential survival and proliferative factor for hematopoietic progenitor cells in all divisions up to maturity. It also stimulates growth in some epithelial cells and osteoclasts. GM-CSF is produced by a variety of cell types (monocytes, endothelial cells, T-cells, fibroblasts, mitogen-stimulated B-cells, and LPS-stimulated macrophages). GM-CSF is secreted as a single chain glycoprotein containing 128 amino acids for human with a conserved disulfide bond. Human and murine GM-CSF share approx. 54% sequence homology and do not cross-react in bioactivity.

Physical form

Lyophilized from 10 mM acetic acid plus 250 μg BSA.

Analysis Note

The EC50 activity of mouse GM-CSF is tested in culture using murine FDP-1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Skoda
Journal of receptor and signal transduction research, 19(1-4), 741-772 (1999-03-11)
The helical cytokines constitute a family of proteins with a common three-dimensional structure. They exert a wide variety of biological effects with a preference for the hematopoietic system. The effects of helical cytokines are mediated by cell surface receptors, which
M A Guthridge et al.
Stem cells (Dayton, Ohio), 16(5), 301-313 (1998-10-10)
The process of ligand binding leading to receptor activation is an ordered and sequential one. High-affinity binding of GM-CSF, interleukin 3 (IL-3), and IL-5 to their receptors induces a number of key events at the cell surface and within the
M H Heim
Journal of receptor and signal transduction research, 19(1-4), 75-120 (1999-03-11)
The Jak-STAT pathway was originally discovered through the study of interferon induced intracellular signal transduction. Meanwhile, a large number of cytokines, hormones and growth factors have been found to activate Jaks and STATs. Jaks (Janus Kinases) are a unique class
Vasiliki Kyrargyri et al.
Glia, 63(4), 549-566 (2014-10-10)
Microglia are CNS resident immune cells and a rich source of neuroactive mediators, but their contribution to physiological brain processes such as synaptic plasticity, learning, and memory is not fully understood. In this study, we used mice with partial depletion
A Kelso
Immunology and cell biology, 76(4), 300-317 (1998-09-02)
Cytokines participate in the induction and effector phases of all immune and inflammatory responses. They are therefore obvious tools and targets for strategies designed to promote, inhibit or redirect these responses. However, the complexity of the cytokine network has hindered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service