Skip to Content
Merck
All Photos(4)

Key Documents

F9048

Sigma-Aldrich

D-(−)-Fructose

98.0-102.0% dry basis, meets USP testing specifications

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

natural (organic)

Agency

USP/NF
meets USP testing specifications

Assay

98.0-102.0% dry basis

form

crystalline

impurities

<0.018% chloride content
<0.025% sulphate

color

colorless

useful pH range

5.0-7 (25 °C, 18 g/L)

mp

119-122 °C (dec.) (lit.)

solubility

water: 790 g/L at 20 °C

cation traces

Ca: ≤0.005%
Mg: ≤0.005%

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Identification of hyperthermophilic D-allulose 3-epimerase from Thermotoga sp. and its application as a high-performance biocatalyst for D-allulose synthesis: This research highlights the application of D-(−)-Fructose in the biocatalytic synthesis of D-allulose, emphasizing its potential in the production of low-calorie sweeteners in pharmaceutical formulations (Shen et al., 2024).

  • Staphylococcus hsinchuensis sp. nov., Isolated from Soymilk: In this study, D-(−)-Fructose is utilized in the media for culturing and identifying new microbial species, underscoring its role in microbiological research within biotechnological and pharmaceutical fields (Wang et al., 2024).

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanli Hua et al.
Translational neuroscience, 12(1), 351-361 (2021-10-28)
Formaldehyde (FA) is a commercially important chemical applied in industry and scientific research. However, FA has a distinct impact on learning and memory. Although the mechanisms of FA toxicity have been well studied, additional research is required to establish the
B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
K Shiraga et al.
Carbohydrate research, 406, 46-54 (2015-02-07)
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on
Marine Zwicke et al.
Annals of botany, 116(6), 1001-1015 (2015-04-09)
Extreme climatic events such as severe droughts are expected to increase with climate change and to limit grassland perennity. The present study aimed to characterize the adaptive responses by which temperate herbaceous grassland species resist, survive and recover from a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service