Skip to Content
Merck
All Photos(2)

Documents

F8020

Sigma-Aldrich

Fish oil from menhaden

crude source of omega-3 fatty acids

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

density

0.93 g/mL at 25 °C (lit.)

lipid type

oils

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Fish oil is derived from the tissues of oily fish. It lowers blood pressure and high triglyceride levels. Fish oil may be used to treat depression, psychosis, attention deficit-hyperactivity disorder and Alzheimer disease. It reduces inflammation, enhances cognition and decreases cardiovascular disease risk.

Application

Fish oil from Menhaden has been used:
  • to prevent lixiviation of the soybean trypsin inhibitor
  • as a dispersant to study the fabrication and laser performance of planar waveguide lutetium aluminium garnet (LuAG) ceramics
  • as a supplement to reduce colonic damage and inflammatory markers

Biochem/physiol Actions

Menhaden Oil is rich in omega-3 fatty acids. When used to partially supplement a high-fat diet in diabetic rats, Menhaden oil improved end points for peripheral neuropathy by improving motor and sensory nerve conduction velocity. Decreased accumulation of fatty acids in rat livers was also demonstrated.

Other Notes

Contains palmitic plus stearic acid (total): ≤ 30.0%
Contains 20.0 to 31.0% omega-3 (octadecatetraenoic, eicosapentaenoic and docosahexaenoic) fatty acids as triglycerides

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litopenaeus vannamei digestive metallo peptidases compensate for anti-nutritional SBTI in feed
Maytorena-Verdugo CI, et al.
Aquaculture (Amsterdam, Netherlands), 473, 508-512 (2017)
Fabrication and laser performance of planar waveguide LuAG/Yb: LuAG/LuAG ceramics
Jiang N, et al.
Optical Materials, 89, 149-156 (2019)
Fish oil supplements in New Zealand are highly oxidised and do not meet label content of n-3 PUFA
Albert BB, et al.
Scientific reports, 5, 7928-7928 (2015)
Fish Oil Supplementation and Athletic Performance
Volpe SL
ACSM?s Health & Fitness Journal , 16(5), 31-32 (2012)
alpha-Linolenic acid (ALA) is an anti-inflammatory agent in inflammatory bowel disease
Reifen R, et al.
The Journal of Nutritional Biochemistry, 26(12), 1632-1640 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service