Skip to Content
Merck
All Photos(3)

Key Documents

F7799

Sigma-Aldrich

Anti-c-Fos Antibody

rabbit polyclonal

Synonym(s):

C-Fos Antibody, C-Fos Antibody - Anti-c-Fos antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

Product Name

Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50-60 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:5,000 using human colorectal carcinoma tissue
western blot: 1:1,000 using nuclear extract of a phorbol ester-treated human A431 cells or nuclear extract of human A431 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

General description

c-Fos belongs to a family of transcription factors, including FosB, Fra1, Fra2. The fos oncogene was isolated as a retroviral transforming gene (v-fos) carried by the FBJ-murine osteosarcoma virus. c-Fos undergoes post-translational modifications and other forms have been described including 57 Da, 60 kDa and 62 kDa proteins. Stable expression of c-Fos has been shown in developing bone tissue and teeth, hematopoietic cells,germ cells, and in the central nervous system. High levels of c-Fos are expressed in human full term fetal membranes.

Immunogen

synthetic peptide corresponding to the N-terminal region of the human c-Fos proto-oncogene (p55), amino acids 3-16.

Application

Rabbit polyclonal anti-c-Fos antibody recognizes c-Fos by immunoblotting, (as single or multiple bands at 50-62 kDa) and by immunohistochemistry. By immunoblotting, the antibody may also detect bands of ~40 kDa representing c-Fos degradation products. Staining of c-Fos by immunoblotting is specifically inhibited with c-Fos immunizing peptide (c-Fos human, amino acids 3-16 with C-terminally added lysine).
Anti-c-Fos antibody produced in rabbit has been used in immunoassays to determine the c-fos immunoreactivity in spinal cord sections. It has been used in Immunohistochemistry.
It may be used as a probe to determine the presence and roles of c-Fos in a wide variety of cell processes including cell proliferation and differentiation, and responses to cell damage or stress.

Biochem/physiol Actions

c-Fos (AP-1 transcription factor subunit) plays an important role in cell proliferation and differentiation. In addition, it is involved in cellular responses to stress, cell damage and death, and has a central role in normal bone and hematopoietic cell development and in oncogenesis. c-Fos is rapidly and transiently induced in almost every cell type upon stimulation by a variety of extracellular stimuli, including stress, mitogenic growth factors, cytokines, neurotransmitters and pharmacological agents. Increased expression of c-Fos is found in some human carcinomas, including colon and pancreatic adenocarcinoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suk Wai Lam et al.
Virchows Archiv : an international journal of pathology, 476(3), 455-463 (2019-11-27)
Osteoid osteoma and osteoblastoma are bone-forming tumors shown to harbor FOS (87%) and FOSB (3%) rearrangements. The aim was to evaluate the immunohistochemical expression of FOS and FOSB in these tumors in comparison to other bone tumors, to evaluate the
Shuo Chen et al.
Nature, 586(7828), 270-274 (2020-10-02)
The ability to recognize information that is incongruous with previous experience is critical for survival. Novelty signals have therefore evolved in the mammalian brain to enhance attention, perception and memory1,2. Although the importance of regions such as the ventral tegmental
Osteoblasts are target cells for transformation in c-fos transgenic mice.
Grigoriadis A E, et al.
The Journal of Cell Biology, 122(3), 685-701 (1993)
Keke Ren et al.
Frontiers in neural circuits, 11, 57-57 (2017-09-02)
As the main input nucleus of the basal ganglion, the striatum executes different functions, including motivation, reward and attention. The functions of the striatum highly rely on its subregions that receive projections from various cortical areas and the distribution of
Toyoshi Umezu et al.
Toxicology reports, 3, 552-562 (2016-06-21)
Bupropion (BUP), a substituted phenyl-ethylamine, has been utilized for the treatment of depression and for smoking cessation, however, one concern is that BUP may increase a risk of psychosis similar to other substituted phenyl-ethylamine amphetamine (AMPH) and methamphetamine (MetAMPH). BUP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service