Skip to Content
Merck

F7050

Fatty Acid Supplement

Animal-component free, liquid, sterile-filtered, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

functional group

carboxylic acid

shipped in

ambient

storage temp.

2-8°C

Application

Recommended for use at 0.25 to 0.5 ml per liter with CHO cells but is considered to have broader application.

Other Notes

Animal-component free. Proprietary aqueous mixture of fatty acids that have been found to be optimal for cell growth and protein expression.


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library