Skip to Content
F6521
Jewelers forceps, Dumont No. 5
L 4 1/4 in. (11 cm), Inox alloy
Synonym(s):
Dumont forceps, Fine-tipped forceps, Precision forceps
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
eCl@ss:
32035504
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
Inox alloy
L
4 1/4 in. (11 cm)
tip W × D
0.05 mm × 0.02 mm , straight
General description
Bevelled, very fine shanks
Analysis Note
Biology grade
Choose from one of the most recent versions:
Certificates of Analysis (COA)