Skip to Content
Merck
All Photos(1)

Documents

F5305

Sigma-Aldrich

Monoclonal Anti-Fibroblast Growth Factor-Basic antibody produced in mouse

clone 10060, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-FGF2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10060, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 2-8 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF2(2247)

Looking for similar products? Visit Product Comparison Guide

General description

Fibroblast Growth Factor Basic (also known as bFGF) is a pleotropic cytokine that that is a potent mitogenic factor. On binding to its cognate receptor tyrosine kinase, FGFR, bFGF is capable of activation of pathways such as MAPK and PI3K and produce cellular effects in varied cell types. Fibroblast Growth Factor-basic is repoted to induce proliferation of fibroblasts, differentiation, wound healing and regeneration, limb formation, tissue remodelling and angiogenesis. The role of bFGF in chondrocyte proliferation and repair has established the role of bFGF in maintaining cartilage and intervertebral disc homeostasis. Along with VEGF, bFGF is important in angiogenesis and tumorigenesis in response to inflammatory cytokines
Monoclonal Anti-Fibroblast Growth Factor Basic recognizes human Fibroblast Growth Factor Basic. It also recognizes bovine bFGF. It shows 0.2% cross-reactivity with bovine FGF acidic. It does not react with human FGF acidic, FGF-4, -6 or -7.

Immunogen

purified, E. coli-derived recombinant human fibroblast growth factor basic.

Application

Monoclonal Anti-Fibroblast Growth Factor Basic may be used as capture antibody in ELISA at a recommended concentration range of 2-8 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service