Skip to Content
Merck

F5178

Flavin-Containing Monooxygenase-5, Microsomes human

recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution (2.5 mg in 100 mM Potassium Phosphate Buffer)

Synonym(s):

FMO-5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥700 units/mg protein
Recombinant:
expressed in baculovirus infected insect cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected insect cells

Quality Segment

form

buffered aqueous solution (2.5 mg in 100 mM Potassium Phosphate Buffer)

specific activity

≥700 units/mg protein

mol wt

55-60 kDa

packaging

vial of 2.5 mg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FMO5(2330)

Application

FMO5 is the only FMO isoform expressed in both adult and fetal human liver.

Biochem/physiol Actions

FMO′s are responsible for catalyzing the oxygenation of a variety of xenobiotic and drug compounds including environmental pollutants and carcinogens, which demonstrate a nucleophilic nitrogen, sulfur, selenium, and phosphorous-containing moiety associated with their respective oxides. These enzymes are classified as Phase I or oxidative drug metabolism enzymes. FMO5 is the only FMO isoform expressed in both adult and fetal human liver.

Physical form

Solution in 100 mM potassium phosphate buffer, pH 7.4.

Other Notes

methyl p-tolyl sulfide oxidase activity: One unit will catalyze the production of 1.0 picomole of methyl p-tolyl sulfoxide.


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library