Skip to Content
Merck
All Photos(1)

Documents

F4639

Sigma-Aldrich

Monoclonal Anti-Fibrinogen antibody produced in mouse

clone FG-21, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

FG-21, monoclonal

contains

15 mM sodium azide

species reactivity

human, baboon

technique(s)

indirect ELISA: 1:4,000

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Fibrinogen, is a thrombin-coagulable soluble plasma 340kDa glycoprotein, composed of paired sets of three subunits i.e. α, β, γ. Among three subunits, β and γ subunits contain one N-glycosylation site, which is occupied by a biantennary N-glycan. It contains three pairs of disulfide bonded chains called α, β, γ which further folded into four structural domains: the D, E, connector and the COOH-terminal region of the Aα chain.
Monoclonal Anti-Fibrinogen (mouse IgG2a isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

The antibody is specific for human fibrinogen and recognizes an epitope sensitive to reduction commonly used in immunoblotting assays. It also reacts with synthetic human fibrinopeptides A and B, fibrinogen related peptides and baboon fibrinogen. No cross-reactivity with bovine, pig, sheep, horse, dog, goat, rabbit, guinea pig, or rat is observed.

Immunogen

purified haptoglobin from pooled human plasma containing the three major haptotypes.

Application

Monoclonal Anti-Fibrinogen antibody produced in mouse has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • fibrinogen adsorption assay

Biochem/physiol Actions

Fibrinogen, a blood coagulation protein, is regarded as the central protein in the blood coagulation system.Fibrinogen plays a crucial role in protecting the vascular network against the loss of blood after tissue injury.
Plasmin attacks the Aα chain COOH domain to produce the heterogeneous fragment X. After a series of degradation, it ended with terminal digestion products: fragments D and E which represent the major globular domains in fibrinogen. Mutations in this gene leads to several disorders including hypofibrinogenemia, afibrinogenemia.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of Apo-A1 as a biomarker for early diagnosis of bladder transitional cell carcinoma
Li H, et al.
Proteome Science, 9(1), 21-21 (2011)
Photon-modulated changes of cell attachments on poly (spiropyran-co-methyl methacrylate) membranes
Higuchi A, et al.
Biomacromolecules, 5(5), 1770-1774 (2004)
Preservation of hematopoietic stem and progenitor cells from umbilical cord blood stored in a surface derivatized with polymer nanosegments
Higuchi A, et al.
Biomacromolecules, 9(2), 634-639 (2008)
N E Kirschbaum et al.
The Journal of biological chemistry, 265(23), 13669-13676 (1990-08-15)
The COOH-terminal portion of the A alpha chain of human fibrinogen is highly susceptible to proteolytic degradation. This property has prevented isolation of the COOH-terminal domain of fibrinogen for the direct investigation of its functional characteristics. Human fibrinogen was degraded
Y Fang et al.
Haemophilia : the official journal of the World Federation of Hemophilia, 12(6), 615-620 (2006-11-07)
Congenital afibrinogenaemia is a rare autosomal recessive disorder, characterized by the complete absence or extremely reduced level of fibrinogen (Fg). We attempted to analyse the phenotype and genotype in two Chinese families with congenital afibrinogenaemia. Coagulation studies including activated partial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service