Skip to Content
Merck

F3388

Ferric citrate

BioReagent, suitable for cell culture

Synonym(s):

Iron(III) citrate, Iron(III) citrate hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H5FeO7
CAS Number:
Molecular Weight:
244.94
NACRES:
NA.75
PubChem Substance ID:
UNSPSC Code:
12352205
EC Number:
222-536-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Level

product line

BioReagent

technique(s)

cell culture | mammalian: suitable

solubility

hot water: 0.01 g/mL, clear, orange-yellow

shipped in

ambient

storage temp.

room temp

SMILES string

OC12CC(=O)O[Fe](OC(=O)C1)OC2=O

InChI

1S/C6H8O7.Fe/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+3/p-3

InChI key

NPFOYSMITVOQOS-UHFFFAOYSA-K

Application

Ferric citrate provides a source of iron used in cell culture applications. It provides iron in a less toxic form than free iron salts. Ferric citrate has been used as medium supplement in human osteosarcoma cell (U2OS) culture, human embryonic kidney (HEK293) cells and adult primary cardiomyocytes.

Biochem/physiol Actions

Ferric citrate is used as a component of protein-free medium. It is used as a substitute for transferrin in mouse hybridomas culture. Ferric citrate enhances IgM production. It indirectly stimulates the mitogen-activated protein kinase (MAPK) by favoring high amphiregulin oncoprotein levels in colon cancer.  Ferric citrate is effective in binding to phosphate in patients with dialysis-dependent chronic kidney disease.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Development of a protein-free medium with iron salts replacing transferrin for a human-human hybridoma
Nagira K, et al.
Bioscience, Biotechnology, and Biochemistry, 59(4), 743-745 (1995)
Nonantibiotic effects of fluoroquinolones in mammalian cells
Badal S, et al.
The Journal of Biological Chemistry, 290(36), 22287-22297 (2015)
Ferric citrate and ferric EDTA but not ferrous sulfate drive amphiregulin-mediated activation of the MAP kinase ERK in gut epithelial cancer cells
Scheers NM, et al.
Oncotarget, 9(24), 17066-17066 (2018)



Global Trade Item Number

SKUGTIN
F3388-250G04061833613849
F3388-1KG04061833613832