Skip to Content
Merck

Skip To

F3385

Fetuin from fetal bovine serum

lyophilized powder, BioReagent, suitable for cell culture

Synonym(s):

Fetuin from fetal calf serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-762-7
NACRES:
NA.75
MDL number:
Form:
lyophilized powder
Mol wt:
48.4 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

lyophilized powder

mol wt

48.4 kDa

technique(s)

cell culture | mammalian: suitable

impurities

≤0.3% free N-acetylneuraminic acid

solubility

H2O: 1 mg/mL

storage temp.

2-8°C

General description

Fetuin from fetal bovine serum is a plasma glycoprotein. It is a 64 kDa glycoprotein, which is produced by the liver and adipose tissue.
A glycoprotein derived from FBS that is used as Fetuin has been used at a concentration of 500 mg/ml to supplement serum free F12 medium (along with insulin, transferrin and 2-mercaptoethanol) in culture of embryonal carcinoma cells.

Application

Fetuin from fetal bovine serum has been used:
  • as a substrate to determine neuraminidase activity
  • to validate the specificity of sialidases
  • to stabilize m-calpain

Biochem/physiol Actions

Fetuin acts as an inhibitor of insulin receptor tyrosine kinase. It modulates bone remodeling and calcium metabolism. Fetuin regulates vascular calcification, insulin resistance, protease activity control, keratinocytes migration and breast tumor cell proliferative signaling. It functions as a biomarker for neurodegenerative diseases. Fetuin exhibits anti-inflammatory property.

Preparation Note

Prepared by ammonium sulfate fractionation of fetal bovine serum by the method of Pederson, K.O., J. Phys. and Colloid Chem., 51, 164 (1947).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2379F3004341506
description

lyophilized powder, BioReagent, suitable for cell culture

description

lyophilized powder, suitable for cell culture

description

lyophilized powder

description

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, MALDI-MS: suitable

technique(s)

cell culture | mammalian: suitable

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized solid

mol wt

48.4 kDa

mol wt

-

mol wt

-

mol wt

-

solubility

H2O: 1 mg/mL

solubility

H2O: 1 mg/mL, colorless to faint yellow or tan

solubility

-

solubility

water: 50 mg/mL

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

impurities

≤0.3% free N-acetylneuraminic acid

impurities

≤0.3% free N-acetylneuraminic acid

impurities

~0.2% free N-acetylneuraminic acid

impurities

-


Still not finding the right product?

Explore all of our products under Fetuin from fetal bovine serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters