Skip to Content
Merck
All Photos(1)

Documents

F1553

Sigma-Aldrich

S-(+)-Fluoxetine hydrochloride

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18NOF3 · HCl
CAS Number:
Molecular Weight:
345.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

optical activity

[α]/D +14 to +19°, c = 1 in chloroform-d

storage condition

desiccated

color

white

solubility

H2O: soluble ≥20 mg/mL

originator

Eli Lilly

storage temp.

room temp

SMILES string

Cl.CNCC[C@H](Oc1ccc(cc1)C(F)(F)F)c2ccccc2

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H/t16-;/m0./s1

InChI key

GIYXAJPCNFJEHY-NTISSMGPSA-N

General description

Fluoxetine hydrochloride or fluoxetine hcl is a psychotropic agent and one of the initial members of the anti-depressant class of drugs known as selective serotonin-reuptake inhibitors (SSRIs). It is the active ingredient of Prozac®.

Biochem/physiol Actions

Fluoxetine hcl (hydrochloride) is a selective serotonin reuptake inhibitor and functions as an antidepressant. This drug works at presynaptic terminals where it prevents the reuptake of serotonin, resulting in the accumulation of serotonin in extracellular fluid at synapses.
Selective serotonin reuptake inhibitor.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Prozac is a registered trademark of Eli Lilly and Co.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hasebe et al.
British journal of pharmacology, 174(4), 314-327 (2016-12-18)
Prefrontal dopamine release by the combined activation of 5-HT Male mice were treated with picrotoxin to decrease GABA Picrotoxin caused an anxiogenic effect on three behavioural tests, but it did not affect the immobility time in the forced swim test.
David T Wong et al.
Nature reviews. Drug discovery, 4(9), 764-774 (2005-08-27)
In the early 1970s, evidence of the role of serotonin (5-hydroxytryptamine or 5-HT) in depression began to emerge and the hypothesis that enhancing 5-HT neurotransmission would be a viable mechanism to mediate antidepressant response was put forward. On the basis
Xin-Xian Ye et al.
Nuclear medicine and biology, 31(5), 557-562 (2004-06-29)
A radiopharmaceutical, (123)I-labeled 2-((2-((dimethylamino)methyl)phenyl)thio)-5-iodophenylamine ([(123)I]ADAM), has been developed recently for evaluation of how serotonin transporters (SERT) function in the brain. However, the detailed biodistribution and specific binding in certain brain areas are not well investigated. In this study, both phosphor
Jing Yuan et al.
Nature chemical biology, 5(10), 765-771 (2009-09-08)
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service