Skip to Content
Merck
All Photos(1)

Key Documents

F112

Sigma-Aldrich

(+)-Fenfluramine hydrochloride

Synonym(s):

(+)-N-Ethyl-α-methyl-m-[trifluoromethyl]phenethylamine hydrochloride, Dexfenfluramine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16F3N · HCl
CAS Number:
Molecular Weight:
267.72
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

powder

drug control

USDEA Schedule IV

color

white to off-white

solubility

H2O: soluble 10 mg/mL

application(s)

forensics and toxicology

storage temp.

room temp

SMILES string

Cl[H].CCN[C@@H](C)Cc1cccc(c1)C(F)(F)F

InChI

1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H/t9-;/m0./s1

InChI key

ZXKXJHAOUFHNAS-FVGYRXGTSA-N

Biochem/physiol Actions

(+)-Fenfluramine is a serotonin uptake inhibitor; anoxexic.
(+)-Fenfluramine is a serotonin uptake inhibitor; anoxexic. (+)-Fenfluramine is neurotoxic on prolonged administration or at high dosage. (+)-Fenfluramine releases serotonin from axon terminals by a nonexocytotic mechanism.

Other Notes

Active isomer

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret R Maclean et al.
Advances in experimental medicine and biology, 661, 309-322 (2010-03-06)
The serotonin hypothesis of pulmonary arterial hypertension (PAH) arose after an outbreak of PAH in patients taking the anorexigenic drugs aminorex and dexfenfluramine. Both of these drugs are serotonin transporter (SERT) substrates and indirect serotinergic agonists. There is now a
Rima Hajjo et al.
Journal of medicinal chemistry, 53(21), 7573-7586 (2010-10-21)
Some antipsychotic drugs are known to cause valvular heart disease by activating serotonin 5-HT(2B) receptors. We have developed and validated binary classification QSAR models capable of predicting potential 5-HT(2B) actives. The classification accuracies of the models built to discriminate 5-HT(2B)
Delphine Natali et al.
Chest, 140(4), 1066-1068 (2011-10-06)
We report a case of pulmonary arterial hypertension (PAH) occurring in a patient with Cowden syndrome with a mutation in the phosphatase and tensin (PTEN) tumor suppressor gene, in the context of exposure to the appetite suppressant dexfenfluramine. Anorexigen exposure
Nicolas Desbuards et al.
European journal of pharmacology, 602(2-3), 355-363 (2008-12-04)
The anorectic drug, dexfenfluramine has been associated with an increase in the relative risk of developing pulmonary hypertension. 5-hydroxytryptamine (5-HT) is a mitogen for smooth muscle cell, an effect that relies on 5-HT transporter expression and which has been proposed
Richard W Foltin
Physiology & behavior, 103(5), 493-500 (2011-04-23)
We examined the effects of periodic access to a palatable, high sugar content food (candy) in 8 male baboons on the anorectic response to d-amphetamine, which increases dopamine, and dexfenfluramine, which increases serotonin. During candy access, up to 200 candies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service