Key Documents
EP5810702003
Eppendorf® MTP-buckets Retrofit kit
Eppendorf(R) MTP-buckets Retrofit kit, for 4 MTPs each or 1 deepwell plate, for swing-bucket rotor A-4-62-MTP or for retrof- fitting of swing-bucket- rotor A-4-62, pkg of 4 ea
About This Item
UNSPSC Code:
41103910
NACRES:
NB.82
feature
Eppendorf(R) MTP-buckets Retrofit kit, for 4 MTPs each or 1 deepwell plate, for swing-bucket rotor A-4-62-MTP or for retrof- fitting of swing-bucket- rotor A-4-62
packaging
pkg of 4 ea
manufacturer/tradename
Eppendorf® 5810702003
technique(s)
centrifugation: suitable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)