About This Item
NACRES:
NB.82
UNSPSC Code:
41103911
Compatibility:
for use with 100 mL rectangular bucket
Manufacturer/tradename:
Eppendorf® 5804753003
feature
holds standard, Vacutainer, Falcon and micro test tubes 3-15 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804753003
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL rectangular bucket
General description
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Eppendorf® Adapter For 100 mL Rectangular Bucket
Choose from one of the most recent versions:
Already Own This Product?