Skip to Content
Merck
All Photos(1)

Key Documents

EP3123000349

Eppendorf® Research plus G, Basic

single-channel, variable, volume 0.25-2.5 mL, red, without tip box

Synonym(s):

Pipettors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121504

feature

adjustable
manual
single-channel variable
without tip box

packaging

case of 1 ea
pack of 1 ea

manufacturer/tradename

Eppendorf® 3123000349

volume

0.25-2.5 mL
1 μL

color

red

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Eppendorf Research plus is one of the safest and most ergonomic mechanical pipettes currently available. It not only meets the highest requirements for precision and accuracy, but also offers a high degree of flexibility thanks to a large selection of variants. This makes the Research plus an indispensable companion in your laboratory. Thanks to its remarkably low weight and reduced effort when pipetting, this micropipette helps to relieve the strain on your hand and arm.

Application

  • Forward pipetting
  • Reverse pipetting
  • Removal of excess material
  • Sample mixing
  • Phase extraction
  • Filling of plates, gels and reaction vessels

Features and Benefits

  • Mechanical air cushion pipette with three-button operation as standard (one control for volume selection, one for pipetting and one for tip ejection)
  • Ideal for accurate, easy and ergonomic pipetting of aqueous solutions by hand
  • Convince yourself of the difference in weight and effort required when pipetting: Developed according to the principles of the Eppendorf PhysioCare Concept
  • Benefit from one of the pipettes with the lowest operating and tip ejection forces on the pipette market to reduce hand and arm strain
  • Minimized attachment forces thanks to a spring-loaded tip cone (available for all pipettes up to 1 mL)
  • Adjust your pipette in seconds to improve accuracy when pipetting difficult liquids such as ethanol or when pipetting at high altitudes. Then return to factory adjustment without calibration
  • Depending on your needs and application, autoclave the entire pipette or just the base to ensure proper decontamination
  • High-quality material ensures high UV and chemical resistance; special single-channel bases available for aggressive chemicals
  • Single-channel pipettes with ACT label and epT.IPS pipette tips help you make an informed choice when it comes to sustainability

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service