Skip to Content
Merck
All Photos(1)

Key Documents

EHU050131

Sigma-Aldrich

MISSION® esiRNA

targeting human RAF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGAGTGCTGTGCAGTGTTCAGACTTCTCCACGAACACAAAGGTAAAAAAGCACGCTTAGATTGGAATACTGATGCTGCGTCTTTGATTGGAGAAGAACTTCAAGTAGATTTCCTGGATCATGTTCCCCTCACAACACACAACTTTGCTCGGAAGACGTTCCTGAAGCTTGCCTTCTGTGACATCTGTCAGAAATTCCTGCTCAATGGATTTCGATGTCAGACTTGTGGCTACAAATTTCATGAGCACTGTAGCACCAAAGTACCTACTATGTGTGTGGACTGGAGTAACATCAGACAACTCTTATTGTTTCCAAATTCCACTATTGGTGATAGTGGAGTCCCAGCACTACCTTCTTTGACTATGCGTCGTATGCGAGAGTCTGTTTCCAGGATGCCTGTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Lin et al.
Cell cycle (Georgetown, Tex.), 19(19), 2496-2508 (2020-09-16)
Since the essential involvement of microRNAs (miRNAs) in the development and progression of GC, the study was for the exploration of the value of microRNA-7 (miR-7) in the evaluation of neoadjuvant chemotherapy for gastric cancer (GC) and its effects on
Xiaoyu Zhang et al.
ACS central science, 4(1), 71-80 (2018-02-03)
The KRAS gene encodes two isoforms, KRas4a and KRas4b. Differences in the signaling functions of the two KRas proteins are poorly understood. Here we report the comparative and nucleotide-dependent interactomes of KRas4a and KRas4b. Many previously unknown interacting proteins were
Ines Jeric et al.
Nature communications, 7, 13781-13781 (2016-12-22)
Hepatocellular carcinoma (HCC) is a leading cause of cancer deaths, but its molecular heterogeneity hampers the design of targeted therapies. Currently, the only therapeutic option for advanced HCC is Sorafenib, an inhibitor whose targets include RAF. Unexpectedly, RAF1 expression is
Hajime Yurugi et al.
Journal of cell science, 133(12) (2020-06-06)
The RAS oncogenes are frequently mutated in human cancers and among the three isoforms (KRAS, HRAS and NRAS), KRAS is the most frequently mutated oncogene. Here, we demonstrate that a subset of flavaglines, a class of natural anti-tumour drugs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service