Skip to Content
Merck

EC1

Escherichia coli

Strain K12, lyophilized cells

Synonym(s):

E. coli K-12

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Escherichia coli, Strain K12, lyophilized cells

sterility

non-sterile (not processed or packaged aseptically)

form

lyophilized cells

suitability

not intended for starter culture

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

General description

Escherichia coli (E.coli) is a non-spore forming, Gram-negative, rod-shaped facultative anerobe, which is found in the human gastrointestinal tract. It belongs to the family of Enterobacteriaceae. E.coli is chemoorganotrophic and grows at 37°C. Pathogenic E.coli strains is associated with diarrhea, septicemia, meningitis and urinary tract infections. It inhibits the colonization of the gut by harmful bacteria. E.coli acts as an indicator of fecal contamination.

Other Notes

A mutant which is rich in alkaline phosphatase.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple antibiotic resistance indexing of Escherichia coli to identify high-risk sources of fecal contamination of foods.
Krumperman PH, et al.
Applied and Environmental Microbiology, 46(1), 165-170 (1983)
Joseph M Autry et al.
Analytical biochemistry, 610, 113965-113965 (2020-09-22)
We have analyzed protein expression and enzyme activity of the sarcoplasmic reticulum Ca2+-transporting ATPase (SERCA) in horse gluteal muscle. Horses exhibit a high incidence of recurrent exertional rhabdomyolysis, with myosolic Ca2+ proposed, but yet to be established, as the underlying
Min-Hui Zhao et al.
Biosensors & bioelectronics, 150, 111865-111865 (2019-11-20)
We develop a low-background electrochemical biosensor for one-step detection of uracil DNA glycosylase (UDG) based on the host-guest interaction and iron-embedded nitrogen-rich carbon nanotube (Fe-N-C) that mimics enzyme-mediated electrocatalysis to achieve signal amplification. In this work, Fe-N-C is initially immobilized
Daria Semenova et al.
New biotechnology, 56, 27-37 (2019-11-11)
Design and development of scale-down approaches, such as microbioreactor (μBR) technologies with integrated sensors, are an adequate solution for rapid, high-throughput and cost-effective screening of valuable reactions and/or production strains, with considerably reduced use of reagents and generation of waste.
D Semenova et al.
The Analyst, 145(3), 1014-1024 (2019-12-13)
The manufacturing of conventional enzymatic biosensors produced via a layer-by-layer (LbL) approach requires expensive instrumentation, and in most cases involves a complex, resource and time-consuming fabrication process. Moreover, LbL assemblies are prone to mechanical instability that leads to irreversible changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service