Skip to Content
Merck
All Photos(3)

Key Documents

E9875

Sigma-Aldrich

Estrone 3-methyl ether

≥97% (HPLC)

Synonym(s):

3-Methoxy-1,3,5(10)-estratrien-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O2
CAS Number:
Molecular Weight:
284.39
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

solubility

chloroform: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

COc1ccc2C3CCC4(C)C(CCC4=O)C3CCc2c1

InChI

1S/C19H24O2/c1-19-10-9-15-14-6-4-13(21-2)11-12(14)3-5-16(15)17(19)7-8-18(19)20/h4,6,11,15-17H,3,5,7-10H2,1-2H3/t15-,16-,17+,19+/m1/s1

InChI key

BCWWDWHFBMPLFQ-VXNCWWDNSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Kozłowska et al.
Scientific reports, 9(1), 10734-10734 (2019-07-26)
Estrone, estradiol, ethynylestradiol and estrone 3-methyl ether underwent a biotransformation process in the submerged culture of Isaria fumosorosea KCh J2. Estrone was transformed into seven metabolites, four of which were glycosylated. Estradiol was selectively glycosylated at C-3 and then transformed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service