Skip to Content
Merck
All Photos(1)

Documents

E9251

Sigma-Aldrich

Ecteola Cellulose

coarse

Synonym(s):

Epichlorohydrin triethanolamine–cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Quality Level

particle size

, coarse

capacity

~0.3 meq/g

Looking for similar products? Visit Product Comparison Guide

Application

Ecteola cellulose is used in affinity chromatography, protein chromatography and heparin applications. Ecteola cellulose has been used in studies to develop methods for evaluating sugar concentrations and to detect inhibition of clot strength in blood samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P E Greilich et al.
Anesthesia and analgesia, 84(1), 31-38 (1997-01-01)
The monoclonal antibody, c7E3 Fab, binds to the platelet surface fibrinogen receptor (GPIIb/IIIa), inhibiting platelet aggregation and clot retraction. We performed an in vitro study to assess the ability of a modification of the thromboelastograph (MTEG) to detect inhibition of
Ana Carolina Campi-Azevedo et al.
Journal of immunological methods, 448, 9-20 (2017-05-18)
Technological innovations in vaccinology have recently contributed to bring about novel insights for the vaccine-induced immune response. While the current protocols that use peripheral blood samples may provide abundant data, a range of distinct components of whole blood samples are
A Calatroni et al.
Clinica chimica acta; international journal of clinical chemistry, 206(3), 167-180 (1992-03-31)
Acid glycosaminoglycans were isolated from normal human plasma: (a) following fractionation of plasma (with protease inhibitors) on Sephadex G-200; (b) by ultrafiltration through membranes with retention of molecules above 50 kDa, with and without previous addition of NaCl, Triton X-100
G M Campo et al.
Journal of chromatography. B, Biomedical sciences and applications, 765(2), 151-160 (2002-01-05)
An improved isocratic high-performance liquid chromatography (HPLC) method for the analysis of L-(-)-fucose. D-(+)-galactosamine, D-(+)-glucosamine, D-(+)-galactose, obtained by hydrolysis of glycosaminoglycans (GAGs) and D-(+)-glucose and D-(+)-mannose is described. The presence in circulation of GAGs, acid polysaccharide sequences of alternate monosaccharide
A M van den Besselaar et al.
Thrombosis and haemostasis, 63(1), 16-23 (1990-02-19)
Five different APTT-reagents, two amidolytic anti-IIa assays, one amidolytic anti-Xa assay, and one coagulometric anti-Xa/anti-IIa assay were used to assess the effect of heparin in patients treated for venous thromboembolic disease. Good correlations were observed between log-transformed APTTs determined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service