Skip to Content
Merck
All Photos(1)

Key Documents

E8909

Sigma-Aldrich

Phosphodiesterase 3B

recombinant, expressed in Sf9 cells

Synonym(s):

3′,5′-cyclic-nucleotide phosphodiesterase, PDE 3B

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Sf9 cells

Quality Level

form

buffered aqueous solution

specific activity

≥1000 units/μg protein

shipped in

dry ice

storage temp.

−70°C

Related Categories

Application

Phosphodiesterase 3B has been used in a study to evaluate the evidence for biological effects of metformin in operable breast cancer. Phosphodiesterase 3B has also been used in a study to investigate the role of intracellular and intercellular communication in perfusion distribution of erythrocyte-derived ATP.

Physical properties

N-terminal GST-tagged 86 kDa protein containing amino acids 592-end

Unit Definition

One unit will convert 1.0 picomole of 3′,5′-cAMP to 5′-AMP per minute at pH 7.4 at 37 °C.

Physical form

Buffered aqueous solution containing Tris-HCl (pH 8.0), NaCl, KCl, Tween-20, DTT, and glycerol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirwan Hadad et al.
Breast cancer research and treatment, 128(3), 783-794 (2011-06-10)
Metformin may reduce the incidence of breast cancer and enhance response to neoadjuvant chemotherapy in diabetic women. This trial examined the effects of metformin on Ki67 and gene expression in primary breast cancer. Non-diabetic women with operable invasive breast cancer
Randy S Sprague et al.
Microcirculation (New York, N.Y. : 1994), 19(5), 430-439 (2012-07-11)
In complex organisms, both intracellular and intercellular communication are critical for the appropriate regulation of the distribution of perfusion to assure optimal O(2) delivery and organ function. The mobile erythrocyte is in a unique position in the circulation as it
Lin Xie et al.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society, 18(4), 444-454 (2011-12-14)
Here we examined whether the expression of a novel immunoregulatory gene set could be used to predict outcomes in murine models of rapamycin-induced cardiac tolerance, spontaneous hepatic tolerance, and cardiac rejection. The expression of the immunoregulatory gene set was assessed
Tamam El-Elimat et al.
Journal of natural products, 76(3), 382-387 (2013-01-11)
Three bioactive compounds were isolated from an organic extract of an ascomycete fungus of the order Chaetothyriales (MSX 47445) using bioactivity-directed fractionation as part of a search for anticancer leads from filamentous fungi. Of these, two were benzoquinones [betulinan A
N B Matveeva et al.
Biofizika, 57(5), 832-839 (2012-11-10)
Possible involvement of autocrine factors into the control of motile behavior via a receptor-mediated mechanism was investigated in Physarum polycephalum plasmodium, a multinuclear amoeboid cell with the auto-oscillatory mode of motility. Cyclic adenosine monophosphate (cAMP) and extracellular cAMP-specific phosphodiesterase, its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service