Skip to Content
Merck
All Photos(1)

Key Documents

E8405

Sigma-Aldrich

EB-47

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N9O6Cl2
CAS Number:
Molecular Weight:
610.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

H2O: soluble 10 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@@H]([C@@H](O)[C@H]3O)C(=O)N4CCN(CC4)CC(=O)Nc5cccc6C(=O)NCc56

InChI

1S/C24H27N9O6/c25-20-16-21(28-10-27-20)33(11-29-16)24-18(36)17(35)19(39-24)23(38)32-6-4-31(5-7-32)9-15(34)30-14-3-1-2-12-13(14)8-26-22(12)37/h1-3,10-11,17-19,24,35-36H,4-9H2,(H,26,37)(H,30,34)(H2,25,27,28)/t17-,18+,19-,24+/m0/s1

InChI key

DDFLFKTXUWPNMV-UAKAABGRSA-N

Application

EB-47 has been used as a poly [ADP-ribose] polymerase 1 (PARP/PARP1/PARPi) inhibitor:
  • to investigate its effect on the activity of matrix metalloproteinase-2 (MMP-2) and compare its inhibitory potencies with the MMP inhibitors
  • in treating human acute monocytic leukemia cells to confirm if PARP1 is responsible for the decrease in high mobility group box protein 1 (HMGB1) supernatant levels
  • to study chromosomal aberrations in response to ionizing radiation (IR) and PARPi in BRCA1-deficient cells that overcome growth arrest (BOGA) cells

Biochem/physiol Actions

1-piperazineacetamide-4-[1-(6-amino-9H-purin-9-yl)-1-deoxy-d-ribofuranuron]-N-(2,3-dihydro-1H-isoindol-4-yl)-1-one (EB-47) can mimic the substrate NAD+ binding. This inhibitor targets the nicotinamide (NI) and adenosine (AD)-subsite within the tankyrase 2 (TNKS2) catalytic domain. EB-47 is one of the most effective TNKS2 inhibitors, with an IC50 of 32 nM, in the presence of piperazine and succinyl linkers that connect the adenosine and isoindolinone cores.
Potent inhibitor of poly (ADP-ribose) polymerase (PARP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teemu Haikarainen et al.
ACS medicinal chemistry letters, 5(1), 18-22 (2014-06-06)
Tankyrases, an enzyme subfamily of human poly(ADP-ribosyl)polymerases, are potential drug targets especially against cancer. We have evaluated inhibition of tankyrases by known PARP inhibitors and report five cocrystal structures of the most potent compounds in complex with human tankyrase 2.
Wei Qiu et al.
Acta crystallographica. Section D, Biological crystallography, 70(Pt 10), 2740-2753 (2014-10-08)
The poly(ADP-ribose) polymerase (PARP) family represents a new class of therapeutic targets with diverse potential disease indications. PARP1 and PARP2 inhibitors have been developed for breast and ovarian tumors manifesting double-stranded DNA-repair defects, whereas tankyrase 1 and 2 (TNKS1 and
Levani Zandarashvili et al.
Science (New York, N.Y.), 368(6486) (2020-04-04)
The success of poly(ADP-ribose) polymerase-1 (PARP-1) inhibitors (PARPi) to treat cancer relates to their ability to trap PARP-1 at the site of a DNA break. Although different forms of PARPi all target the catalytic center of the enzyme, they have
David A Grotsky et al.
The Journal of cell biology, 200(2), 187-202 (2013-01-23)
Loss of 53BP1 rescues BRCA1 deficiency and is associated with BRCA1-deficient and triple-negative breast cancers (TNBC) and with resistance to genotoxic drugs. The mechanisms responsible for decreased 53BP1 transcript and protein levels in tumors remain unknown. Here, we demonstrate that
Adrian C Nicolescu et al.
Biochemical and biophysical research communications, 387(4), 646-650 (2009-07-22)
Matrix metalloproteinase-2 (MMP-2), a ubiquitously expressed zinc-dependent endopeptidase, and poly(ADP-ribosyl) polymerase (PARP), a nuclear enzyme regulating DNA repair, are activated by nitroxidative stress associated with various pathologies. As MMP-2 plays a detrimental role in heart injuries resulting from enhanced nitroxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service