Skip to Content
Merck
All Photos(2)

Key Documents

E6412

Sigma-Aldrich

Cellobiohydrolase I from Hypocrea jecorina

greener alternative

0.13 U/mg, recombinant, expressed in corn

Synonym(s):

Cel7A, Cellobiosidase, Cellulase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in corn

form

liquid

specific activity

0.13 U/mg

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

dry ice

storage temp.

−20°C

General description

Cellubiohydrolase I is an enzyme present in many fungi, but particularly wood rot fungi. It is a monomer of 53 kDa with a catalytic domain and a cellulose binding domain. The reaction adds water to the glucose bonds in cellulose (non-reducing ends of the chain), yielding cellobiose.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Cellobiohydrolase I can be used in combination with endocellulases and b-glucosidase to produce glucose from cellulose.

Biochem/physiol Actions

Cellobiohydrolase (CBH) is a cellulase which degrades cellulose by hydrolysing the 1,4-β-D-glycosidic bonds. CBH is an exocellulase which cleaves two to four units from the ends of cellulose. CBH I cleaves progressively from the reducing end. CBH I is commonly used in detergents for cleaning textiles. Its ezymatic activity ranges from 37° C to 50° C, with its optimal temperature being approximately 45° C. The optimum pH for the enzyme is 5-6.

Unit Definition

Unit Definition: A unit will turn over 1 nmole of methyl-umbelliferyl beta-D cellobioside per min at pH 5 at 50° C.

Physical form

Provided as an ammonium sulfate precipitate with the source as recombinant maize.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolaj Cruys-Bagger et al.
Biotechnology and bioengineering, 109(12), 3199-3204 (2012-07-07)
An amperometric enzyme biosensor for continuous detection of cellobiose has been implemented as an enzyme assay for cellulases. We show that the initial kinetics for cellobiohydrolase I, Cel7A from Trichoderma reesei, acting on different types of cellulose substrates, semi-crystalline and
Zachary K Haviland et al.
The Journal of biological chemistry, 297(3), 101029-101029 (2021-08-03)
Understanding the mechanism by which cellulases from bacteria, fungi, and protozoans catalyze the digestion of lignocellulose is important for developing cost-effective strategies for bioethanol production. Cel7A from the fungus Trichoderma reesei is a model exoglucanase that degrades cellulose strands from
Oren Yaniv et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 7), 819-828 (2012-07-04)
The crystal structure of the family 3b carbohydrate-binding module (CBM3b) of the cellulosomal multimodular hydrolytic enzyme cellobiohydrolase 9A (Cbh9A) from Clostridium thermocellum has been determined. Cbh9A CBM3b crystallized in space group P4(1) with four molecules in the asymmetric unit and
Kiyohiko Igarashi et al.
Methods in enzymology, 510, 169-182 (2012-05-23)
Cellulases hydrolyze β-1,4-glucosidic linkages of insoluble cellulose at the solid/liquid interface, generating soluble cellooligosaccharides. We describe here our method for real-time observation of the behavior of cellulase molecules on the substrate, using high-speed atomic force microscopy (HS-AFM). When glycoside hydrolase
Microbial cellulases-Production, applications and challenges.
Sukumaran RK, et al.
J. Sci. Ind. Res., 64(11), 832-844 (2005)

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service