Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
EC Number:
208-937-9
MDL number:
Assay:
≥93% (HPLC)
Form:
powder
Product Name
16-Epiestriol, ≥93% (HPLC)
Quality Segment
assay
≥93% (HPLC)
form
powder
solubility
ethanol: 0.98-1.02 mg/mL, clear, colorless to faintly yellow
shipped in
ambient
storage temp.
room temp
SMILES string
CC12CCC3C(CCc4cc(O)ccc34)C1CC(O)C2O
InChI
1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3
InChI key
PROQIPRRNZUXQM-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
signalword
Warning
hcodes
Hazard Classifications
Repr. 2 - Skin Irrit. 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

