Skip to Content
Merck

E2375

Emetine dihydrochloride

RNA polymerase inhibitor

Synonym(s):

6′,7′,10,11-Tetramethoxyemetan dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H40N2O4 · 2HCl
CAS Number:
Molecular Weight:
553.56
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Emetine dihydrochloride,

Quality Segment

antibiotic activity spectrum

parasites

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C[C@@]3([H])NCCC4=C3C=C(OC)C(OC)=C4)[C@@H](CC)CN1CCC5=C2C=C(OC)C(OC)=C5

InChI

1S/C29H40N2O4.2ClH/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24;;/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3;2*1H/t18-,21-,24+,25-;;/m0../s1

InChI key

JROGBPMEKVAPEH-GXGBFOEMSA-N

Application

Emetine dihydrochloride has been used:
  • as a protein synthesis inhibitor to study its effects on human papillomavirus type 8 E2 protein half-life
  • to study its effects on the stress granules assembly
  • as a chain-elongation inhibitor in puromycin assay for protein synthesis

Biochem/physiol Actions

Emetine dihydrochloride is a member of the ipecac alkaloids family. It can mediate inhibition of protein and nucleic acid synthesis. It exhibits antiviral activity against dengue virus infection and severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). Emetine dihydrochloride exhibits anti-proliferative effects against bladder cancer cells. It also possesses anti-protozoal activity.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Still not finding the right product?

Explore all of our products under Emetine dihydrochloride


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library